EMAIL THIS PAGE TO A FRIEND
Oncotarget
20(S)-Ginsenoside Rg3 is a novel inhibitor of autophagy and sensitizes hepatocellular carcinoma to doxorubicin.
PMID 24970805
Abstract
Hepatocellular carcinoma (HCC) is the third leading cause of cancer-related deaths worldwide. High mortality from HCC is mainly due to widespread prevalence and the lack of effective treatment, since systemic chemotherapy is ineffective, while the targeted agent Sorafenib extends median survival only briefly. The steroidal saponin 20(S)-ginsenoside Rg3 from Panax ginseng C.A. Meyer is proposed to chemosensitize to various therapeutic drugs through an unknown mechanism. Since autophagy often serves as cell survival mechanism in cancer cells exposed to chemotherapeutic agents, we examined the ability of Rg3 to inhibit autophagy and chemosensitize HCC cell lines to doxorubicin in vitro. We show that Rg3 inhibits late stage autophagy, possibly through changes in gene expression. Doxorubicin-induced autophagy plays a protective role in HCC cells, and therefore Rg3 treatment synergizes with doxorubicin to kill HCC cell lines, but the combination is relatively nontoxic in normal liver cells. In addition, Rg3 was well-tolerated in mice and synergized with doxorubicin to inhibit tumor growth in HCC xenografts in vivo. Since novel in vivo inhibitors of autophagy are desirable for clinical use, we propose that Rg3 is such a compound, and that combination therapy with classical chemotherapeutic drugs may represent an effective therapeutic strategy for HCC treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405458
Anti-ANXA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1305642
ANTI-APG5 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
A0856
Anti-ATG5 (N-terminal) antibody produced in rabbit, affinity isolated antibody, PBS solution
GW22210F
Anti-ATG5 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500448
Anti-ATG5 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA042973
Anti-ATG5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3611
Anti-Beclin-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3613
Anti-Beclin-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103299
Anti-BECN1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2701263
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107220
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody
SAB2105017
Anti-DCTN4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW22005A Anti-GTF2H1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22005B Anti-GTF2H1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV100811
Anti-GTF2H1 antibody produced in rabbit, IgG fraction of antiserum
SAB2104463
Anti-GTF2H1 antibody produced in rabbit, affinity isolated antibody
SAB1408113
Anti-MAP1LC3A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
GW22431A Anti-NUP62 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA005435
Anti-NUP62 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101664
Anti-NUP62 antibody produced in rabbit, affinity isolated antibody
SAB1410438
Anti-NUP62 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
46401
Cycloheximide, PESTANAL®, analytical standard
C15H23NO4
18079
Cycloheximide solution, 0.1%, for microbiology
C15H23NO4
WH0000306M12
Monoclonal Anti-ANXA3 antibody produced in mouse, clone 4F1, purified immunoglobulin, buffered aqueous solution
SAB1403553
Monoclonal Anti-ANXA3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB4100143
Monoclonal Anti-ANXA3 antibody produced in mouse, clone ANXA3-A107, culture supernatant
WH0009474M1
Monoclonal Anti-ATG5 antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution
A2859
Monoclonal Anti-ATG5 antibody produced in mouse, clone ATG5-18, purified immunoglobulin, buffered aqueous solution
SAB4100184
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-B129, culture supernatant
SAB4100208
Monoclonal Anti-BECN1 antibody produced in mouse, clone BECN1-A129, culture supernatant