EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Preclinical evaluation of a high-affinity 18F-trifluoroborate octreotate derivative for somatostatin receptor imaging.
PMID 24970911
Abstract
Recent studies have highlighted the high sensitivity of PET imaging with (68)Ga-labeled octreotide derivatives for the detection and staging of neuroendocrine tumors. A somatostatin receptor ligand that is easily radiolabeled with (18)F-fluoride could improve the availability of PET imaging of neuroendocrine tumors. We report an alkyltrifluoroborate-octreotate conjugate that is radiolabeled in a 1-step (18)F exchange reaction in high yield and with high specific activity. We conjugated a new alkyltrifluoroborate to octreotate to obtain AMBF3-TATE, which was stored in 50-nmol aliquots for radiolabeling. (18)F labeling was performed by (18)F-(19)F isotope exchange with (18)F-fluoride, and the tracer was purified by C18 cartridge separation. The radiochemical yield was 20%-25%. PET imaging and biodistribution were performed on mice bearing AR42J tumor xenografts. AMBF3-TATE bound the somatostatin receptor subtype 2 with high affinity (inhibition constant, 0.13 ± 0.03 nM). Starting with 29.6-37 GBq (0.8-1 Ci) of (18)F-fluoride, more than 7.4 GBq (>200 mCi) of (18)F-AMBF3-TATE were obtained in 25 min (n = 5) with greater than 99% radiochemical purity at high specific activity (>111 GBq [3 Ci]/μmol). (18)F-AMBF3-TATE was stable in plasma. PET imaging and biodistribution showed rapid renal excretion with low liver activity. High tumor uptake (10.11% ± 1.67% injected dose/g, n = 5) was detected at 60 min after injection. Bone uptake was negligible. Tumor-to-liver, tumor-to-blood, tumor-to-muscle, and tumor-to-bone ratios (at 60 min) were 26.2 ± 0.8, 25.1 ± 1.0, 89.0 ± 3.1, and 21.3 ± 3.6, respectively. (18)F-AMBF3-TATE was radiolabeled in high yield and at high specific activity, did not require high-performance liquid chromatography purification, exhibited unexpectedly high binding affinity to somatostatin receptor subtype 2, and showed excellent pharmacokinetic properties in vivo, with high tumor uptake and high contrast ratios.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47378
Fmoc-D-Phe-OH, ≥98.0%
C24H21NO4