EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Inhibition of Akt kinase activity suppresses entry and replication of influenza virus.
PMID 24971535
Abstract
The possibility of the pandemic spread of influenza viruses highlights the need for an effective cure for this life-threatening disease. Influenza A virus, belonging to a family of orthomyxoviruses, is a negative-strand RNA virus which encodes 11 viral proteins. A numbers of intracellular signaling pathways in the host cells interact with influenza the viral proteins, which affect various stages of viral infection and replication. In this study, we investigated how inhibition of Akt kinase activity impacts on influenza virus infection by using "Akt-in", a peptide Akt inhibitor. In PR8 influenza-infected A549 cells, Akt interacted with the NS1 (Non structural protein 1), and hence increased phosphorylation of Akt kinase activity and NS1. Treatment of cells with either "TCL1- or TCL1b-based Akt-in" efficiently suppressed Akt kinase activity while decreasing the levels of phosphorylated NS1; this, in turn, inhibited viral replication in a dose- and time-dependent manner. The inhibitory effect on viral replication appears to not be due to inhibition of the production of inflammatory cytokines, including IL-6 and IL-8, in the host cells. Inhibition of Akt kinase activity in the host cells inhibited the efficiency of viral entry, which is associated with decreased levels of phosphorylated glycogen synthase kinase 3, a substrate of Akt. Thus inhibition of Akt kinase activity in host cells may have therapeutic advantages for influenza virus infection by inhibiting viral entry and replication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22775 Anti-AKT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4503853
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
SAB4504016
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4504015
Anti-phospho-Akt (pSer246) antibody produced in rabbit, affinity isolated antibody
SAB4504110
Anti-phospho-Akt (pTyr326) antibody produced in rabbit, affinity isolated antibody
SAB4504014
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
SAB4300043
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504749
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4503852
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody
SAB4300260
Anti-phospho-AKT1 (pThr450) antibody produced in rabbit, affinity isolated antibody
WH0000207M3
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
SAB1400005
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6D6, purified immunoglobulin, buffered aqueous solution
SAB1400006
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
SAB1400007
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
SAB1400008
Monoclonal Anti-AKT1 antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution