EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
Knockdown of mineralocorticoid or angiotensin II type 1 receptor gene expression in the paraventricular nucleus prevents angiotensin II hypertension in rats.
PMID 24973408
Abstract
Circulating Ang II activates an aldosterone-mineralocorticoid receptor (MR) - angiotensin II (Ang II) - angiotensin type 1 receptor (AT1R) pathway in the hypothalamus. To obtain insights into the actual neuronal projections involved, adeno-associated virus carrying small interfering RNA against either AT1aR (AAV-AT1aR-siRNA) or MR (AAV-MR-siRNA) were infused into the paraventricular nucleus (PVN) in Wistar rats. Intra-PVN infusion of AAV-AT1aR-siRNA or AAV-MR-siRNA decreased AT1R or MR expression in the PVN but not in the subfornical organ (SFO) or supraoptic nucleus (SON). Subcutaneous infusion of Ang II at 500xa0ngxa0kg(-1)xa0min(-1) for 2xa0weeks increased mean arterial pressure by 60-70xa0mmHg, and increased AT1R and MR expression in the SFO, SON and PVN. Intra-PVN AT1aR-siRNA prevented the Ang II-induced increase in AT1R but not MR expression in the PVN, and MR-siRNA prevented MR but not AT1R expression in the PVN. The increases in AT1R and MR expression in both the SFO and the SON were not changed by the two AAV-siRNAs. Specific knockdown of AT1R or MR in the PVN by AAV-siRNA each prevented most of the Ang II-induced hypertension. Prevention of the subcutaneous Ang II-induced increase in MR but not the increase in AT1R by knockdown of MR and vice versa suggests an independent regulation of MR and AT1R expression in the PVN. Both AT1R and MR activation in the PVN play a critical role in Ang II-induced hypertension in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003596
Anti-AGTR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100073
Anti-AGTR1 antibody produced in rabbit, affinity isolated antibody
SAB3500209
Anti-AGTR1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1407767
Anti-AGTRAP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA044120
Anti-AGTRAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411125
Anti-AGTRAP antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA004114
Anti-MRC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA045134
Anti-MRC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1301855 ANTI-MRC1L1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306983
ANTI-MTNR1A (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700731 Anti-MTNR1A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV45599
Anti-NR3C2 antibody produced in rabbit, IgG fraction of antiserum
SAB2104689
Anti-NR3C2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
40718
Diethyl pyrocarbonate, 99% (NT)
C6H10O5
D5758
Diethyl pyrocarbonate, ≥97% (NMR)
C6H10O5
32490
Diethyl pyrocarbonate, purum, ≥97.0% (NT)
C6H10O5
159220
Diethyl pyrocarbonate, 97%
C6H10O5
WH0057085M2
Monoclonal Anti-AGTRAP antibody produced in mouse, clone 1G2, purified immunoglobulin, buffered aqueous solution