EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Preparation and first biological evaluation of novel Re-188/Tc-99m peptide conjugates with substance-P.
PMID 24973465
Abstract
New (188)Re and (99m)Tc peptide conjugates with substance- P (SP) were prepared and biologically evaluated. The radiopharmaceuticals have been labelled with the [M≡N](2+) (M=(99m)Tc, (188)Re) core using a combination of π-donor tridentate and π-acceptor monodentate ancillary ligands. The new radiopharmaceuticals have been prepared through a two-step reaction by simultaneous addition of the tridentate and monodentate ligands to a vial containing a preformed [M≡N](2+) core. The tridentate ligand was formed by linking two cysteine residues to the terminal arginine of the undecapeptide SP, whereas the monodentate ligand was a tertiary phosphine. The preparation of the corresponding Re-188 derivative required developing a more complex chemical procedure to obtain the [Re≡N](2+) core in satisfactory yields. Characterization of the resulting products was obtained by chromatographic methods. Biological evaluation was performed for both Tc-99m and Re-188 derivatives by in-vitro studies on isolated cells expressing NK1-receptors. In-vivo imaging in mice was carried out using a small-animal YAP(S)PET tomograph. New Tc-99m and Re-188 peptide radiopharmaceuticals with SP have been prepared in high-yield and with high-specific activity. Both Tc-99m and Re-188 peptide radioconjugates exhibit high affinity for NK1 receptors, thus giving further evidence to the empirical rule that structurally related Tc-99m and Re-188 radiopharmaceuticals exhibit identical biological properties.