EMAIL THIS PAGE TO A FRIEND
Exercise immunology review
Evaluation of serum leaking enzymes and investigation into new biomarkers for exercise-induced muscle damage.
PMID 24974720
Abstract
This investigation determined whether existing muscle damage markers and organ damage markers respond to an acute eccentric exercise protocol and are associated with affected muscle symptoms. Nine healthy-young men completed one-leg calf-raise exercise with their right leg on a force plate. They performed 10 sets of 40 repetitions of exercise at 0.5 Hz with a load corresponding to half of their body weight, with 3 min rest between sets. The tenderness of medial gastrocnemius, lateral gastrocnemius and soleus, and the ankle active range of motion (ROM) were assessed before, immediately after, 24 h and 48 h, 72 h, 96 h and 168 h after exercise. Blood and urine were collected pre-exercise and 2 h, 4 h, 24 h, 48 h, 72 h and 96 h post-exercise. Serum was analyzed for creatine kinase (CK), aspartate aminotransferase (AST), alanine aminotransferase (ALT), lactate dehydrogenase (LDH) and aldolase (ALD) activities. We also determined heart-type fatty acid-binding protein (H-FABP), intestinal-type fatty acid-binding protein (I-FABP) and liver-type fatty acid-binding protein (L-FABP), neutrophil gelatinase-associated lipocalin (NGAL), interleukin (IL)-17A, IL-23, nerve growth factor (NGF), soluble-Endothelial (sE)-selectin, s-Leukocyte (L)-selectin, s-Platelets (P)-selectin, and 8-isoprostane in plasma and urine. The tenderness of proximal and middle gastrocnemius increased significantly 72 h (p < 0.05, p < 0.01) after exercise. Ankle active ROM in dorsal flexion decreased significantly 48 h (p < 0.05) and 72 h (p < 0.01) after exercise. CK and ALD activities significantly increased at 72 h (p < 0.05) and remained elevated at 96 h (p < 0.01) postexercise compared to pre-exercise values. Also, ALD which showed relatively lower interindividual variability was significantly correlated with tenderness of middle gastrocnemius at 72 h. LDH activity significantly increased 96 h postexercise (p < 0.01), whereas the increase in AST and ALT activities 96 h post-exercise was not significantly different from pre-exercise values. There were no significant changes in FABPs, NGAL, IL-17A, IL-23, NGF, selectins and 8-isoprostanes in plasma and urine. In conclusion, calf-raise exercise induced severe local muscle damage symptoms which were accompanied by increases in both serum CK and ALD activities, but we could not detect any changes in examined markers of organ damage, inflammation and oxidative stress. Further research is needed to determine other more sensitive biomarkers and the underlying mechanisms of exercise-induced muscle damage.