EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
Ewing sarcoma inhibition by disruption of EWSR1-FLI1 transcriptional activity and reactivation of p53.
PMID 24974828
Abstract
Translocations involving ETS-transcription factors, most commonly leading to the EWSR1-FLI1 fusion protein, are the hallmark of Ewing sarcoma. Despite knowledge of this driving molecular event, an effective therapeutic strategy is lacking. To test potential treatment regimes, we established a novel Ewing sarcoma zebrafish engraftment model allowing time-effective, dynamic quantification of Ewing sarcoma progression and tumour burden in vivo, applicable for screening of single and combined compounds. In Ewing sarcoma the tumour-suppressor gene TP53 is commonly found to be wild-type, thus providing an attractive target for treatment. Here, we study TP53 wild-type (EW7, CADO-ES1 and TC32) and TP53-deleted (SK-N-MC) Ewing sarcoma cell lines to investigate the potentiating effect of p53 reactivation by Nutlin-3 on treatment with YK-4-279 to block transcriptional activity of EWSR1-FLI1 protein. Blocking EWSR1-FLI1 transcriptional activity reduced Ewing sarcoma tumour cell burden irrespective of TP53 status. We show that simultaneous YK-4-279 treatment with Nutlin-3 to stabilize p53 resulted in an additive inhibition of TP53 wild-type Ewing sarcoma cell burden, whilst not affecting TP53-deleted Ewing sarcoma cells. Improved inhibition of proliferation and migration by combinatorial treatment was confirmed in vivo by zebrafish engraftments. Mechanistically, both compounds together additively induced apoptosis of tumour cells in vivo by engaging distinct pathways. We propose reactivation of the p53 pathway in combination with complementary targeted therapy by EWSR1-FLI1 transcriptional activity disruption as a valuable strategy against p53 wild-type Ewing sarcoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2105613
Anti-D4S234E, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2101297
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
HPA049830
Anti-KRAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107115
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
SAB2700181
Anti-NS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2502090
Anti-Nsg1 (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA035775
Anti-NSG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501190
Anti-NSG1 antibody produced in rabbit, affinity isolated antibody
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody
P7605 Anti-PARP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33754
Anti-PARP1 antibody produced in rabbit, IgG fraction of antiserum
HPA045168
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503030
Anti-TCTP antibody produced in rabbit, affinity isolated antibody
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300367
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300368
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
SAB4300362
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody
SAB4300363
Anti-TP53 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2102511
Anti-TP53 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102512
Anti-TP53 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2102513
Anti-TP53 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2102514
Anti-TP53 (ab4) antibody produced in rabbit, affinity isolated antibody
SAB2102515
Anti-TP53 (ab5) antibody produced in rabbit, affinity isolated antibody