EMAIL THIS PAGE TO A FRIEND
International immunopharmacology
Airborne pollutant ROFA enhances the allergic airway inflammation through direct modulation of dendritic cells in an uptake-dependent mechanism.
PMID 24975839
Abstract
Studies suggest that airborne pollutants are important cofactors in the exacerbation of lung diseases. The role of DC on the exacerbation of lung inflammation induced by particulate matter pollutants is unclear. We evaluated the effects of residual oil fly ash (ROFA) on the phenotype and function of bone marrow-derived dendritic cells (BMDCs) in vitro and lung dendritic cells (DCs) in vivo, and the subsequent T-cell response. In a model of asthma, exposure to ROFA exacerbated pulmonary inflammation, which was attributed to the increase of eosinophils, IL-5- and IFN-γ-producing T cells, and goblet cells as well as decreased number of Treg and pDC. However, the ROFA showed no ability to modulate the production of anaphylactic IgE. In vitro studies showed that ROFA directly induced the maturation of DCs up-regulating the expression of co-stimulatory molecules and cytokines and MMP production in an uptake-dependent and oxidative stress-dependent manner. Furthermore, ROFA-pulsed BMDC transferred to allergic mice exacerbated eosinophilic inflammation as well as promoted increased epithelial and goblet cells changes. Thus, pollutants may constitute an important and risk factor in the exacerbation of asthma with inhibition of the negative regulatory signals in the lung, with enhanced mDC activation that sustains the recruitment of effector T lymphocytes and eosinophil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22573F Anti-BB1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22783 Anti-CD14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500215
Anti-CD14 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001887
Anti-CD14 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002127
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41929
Anti-CD40 antibody produced in rabbit, affinity isolated antibody
HPA031567
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031568
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103449
Anti-CD40, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA000440
Anti-CD45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501656
Anti-CD80 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405584
Anti-CD80 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411654
Anti-CD86 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1407427
Anti-FOXP3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410887
Anti-FOXP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1400126
Anti-HLA-E antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401182
Anti-HLA-E antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA031454
Anti-HLA-E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1303995
ANTI-KY (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA036492
Anti-KY antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21465 Anti-PTPRC antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22787 Anti-TLR2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
PRS3135
Anti-TLR2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1412080
ANTI-TLR4 antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
SAB1412548
ANTI-TLR4 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1412602
ANTI-TLR4 antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
PRS3141
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500765
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody