EMAIL THIS PAGE TO A FRIEND
IUBMB life
Effects of Smad3 on the proliferation and steroidogenesis in human ovarian luteinized granulosa cells.
PMID 24979474
Abstract
Granulosa cells (GCs) are essential for proper oocyte, follicular development, and steroidogenesis in the ovary. Transforming growth factor β (TGF-β) superfamily members are critical in regulating GCs growth and differentiation. Smad3 is known to serve as a signaling intermediate for the TGF-β; however, the functions of Smad3 in the human GCs remain unidentified. In this study, the luteinized GCs collected from follicular aspirates from patients undergoing in vitro fertilization were cultured and engineered to overexpress and knockdown Smad3, which were validated by RT-PCR and Western blotting. Immunocytochemistry showed that Smad3 protein was strongly expressed in human ovarian luteinized GCs. EdU incorporation demonstrated that Smad3 promoted the proliferation of GCs, and the expression of PCNA was also enhanced by Smad3. ELISA analysis indicated that the secretion of both estradiol and progesterone was stimulated by Smad3. In addition, Smad3 upregulated the level of follicle-stimulating hormone receptor (FSHR), luteinizing hormone receptor (LHR), and protein kinase A (PKA) proteins. We subsequently added special PKA inhibitor H89 into the GCs and found that the stimulating effect on the growth of GCs by Smad3 was blocked partly. The morphology of cultured GCs was changed by Smad3, and the expression level of integrin β1 was enhanced by Smad3. Kindlin-2, an important cellular mediating molecule of integrin β signaling, was expressed in human ovarian luteinized GCs and was upregulated by Smad3. Our results indicated that Smad3 promoted the proliferation and steroidogenesis of human ovarian luteinized GCs, and these effects may be mediated by the FSHR/LHR-PKA signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA040505
Anti-FERMT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
F3929 Anti-Follicle-Stimulating Hormone Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501041
Anti-FSHR antibody produced in rabbit, affinity isolated antibody
SAB4501042
Anti-FSHR antibody produced in rabbit, affinity isolated antibody
K3269
Anti-Kindlin-2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
L6792 Anti-Luteinizing Hormone/Choriogonadotropin Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1401306
Anti-PRKACA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4200318
Anti-PRKACA antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2106172
Anti-PRKACA, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300564
Anti-SMAD3 (Ab-208) antibody produced in rabbit, affinity isolated antibody
SAB4300565
Anti-SMAD3 (Ab-425) antibody produced in rabbit, affinity isolated antibody
AV100621
Anti-SMAD3 antibody produced in rabbit, IgG fraction of antiserum
SAB4200342
Anti-SMAD3 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2702052
Anti-SMAD3 antibody produced in rabbit
SAB4501811
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4501812
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4501813
Anti-SMAD3 antibody produced in rabbit, affinity isolated antibody
SAB4200525
Monoclonal Anti-Kindlin-2 antibody produced in mouse, ~1.0 mg/mL, clone 3A3, purified immunoglobulin
WH0010979M9 Monoclonal Anti-PLEKHC1 antibody produced in mouse, clone 2G11, purified immunoglobulin, buffered aqueous solution
SAB1404238
Monoclonal Anti-PRKACA, (N-terminal) antibody produced in mouse, clone 1D7, purified immunoglobulin, buffered aqueous solution
WH0004088M1
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
WH0004088M5 Monoclonal Anti-SMAD3 antibody produced in mouse, clone 4D5, purified immunoglobulin, buffered aqueous solution
SAB1400160
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 7F3, purified immunoglobulin, buffered aqueous solution
SAB1404037
Monoclonal Anti-SMAD3 antibody produced in mouse, clone 4F9, purified immunoglobulin, buffered aqueous solution