EMAIL THIS PAGE TO A FRIEND
International journal of medical microbiology : IJMM
Metabolic and transcriptional activities of Staphylococcus aureus challenged with high-doses of daptomycin.
PMID 24980509
Abstract
Treatment of stationary growth phase Staphylococcus aureus SA113 with 100-fold of the MIC of the lipopeptide antibiotic daptomycin leaves alive a small fraction of drug tolerant albeit genetically susceptible bacteria. This study shows that cells of this subpopulation exhibit active metabolism even hours after the onset of the drug challenge. Isotopologue profiling using fully (13)C-labeled glucose revealed de novo biosynthesis of the amino acids Ala, Asp, Glu, Ser, Gly and His. The isotopologue composition in Asp and Glu suggested an increased activity of the TCA cycle under daptomycin treatment compared to unaffected stationary growth phase cells. Microarray analysis showed differential expression of specific genes 10 min and 3 h after addition of the drug. Besides factors involved in drug response, a number of metabolic genes appear to shape the signature of daptomycin-tolerant S. aureus cells. These observations will be useful toward the development of new strategies against persisters and related forms of bacterial cells with downshifted physiology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69460
N-Methyltrifluoroacetamide, purum, ≥98.0% (GC)
C3H4F3NO
242098
N-Methyltrifluoroacetamide, 98%
C3H4F3NO