EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
LPS induces the degradation of programmed cell death protein 4 (PDCD4) to release Twist2, activating c-Maf transcription to promote interleukin-10 production.
PMID 24982420
Abstract
Programmed cell death protein 4 (PDCD4) is a tumor suppressor and has also been shown to suppress production of the immunomodulatory cytokine IL-10. The precise role of PDCD4 in IL-10 induction in macrophages is still not fully understood. Incubation of macrophages with inhibitors of PI3K and mTOR blocked LPS-stimulated PDCD4 degradation and expression of c-Maf and IL-10 production. PDCD4 and the transcription factor Twist2 were shown to form a complex in untreated cells. LPS disrupted the complex allowing Twist2 to bind to the c-Maf promoter. PI3K and mTOR inhibitors prevented this disruption by stabilizing PDCD4 and thereby decreased Twist2 binding to the c-Maf promoter and induction of c-Maf mRNA. These results indicate a regulatory role for PDCD4 and Twist2 in LPS-induced IL-10 production in macrophages. LPS promotes PDCD4 degradation via a pathway involving PI3K and mTOR, releasing Twist2, which induces IL-10 via c-Maf.