EMAIL THIS PAGE TO A FRIEND
FEBS letters
Transcriptional activation of OCT4 by the ETS transcription factor PEA3 in NCCIT human embryonic carcinoma cells.
PMID 24983502
Abstract
We examined the molecular mechanism of OCT4 gene regulation by polyomavirus enhancer activator 3 (PEA3) in NCCIT cells. Endogenous PEA3 and OCT4 were significantly elevated in undifferentiated cells and reduced upon differentiation. PEA3 knockdown led to a reduction in OCT4 levels. OCT4 promoter activity was significantly up-regulated by dose-dependent PEA3 overexpression. Deletion and site-directed mutagenesis of the OCT4 promoter revealed a putative binding site within the conserved region 2 (CR2). PEA3 interacted with the binding element within CR2 in NCCIT cells. This study reveals the molecular details of the mechanism by which the oncogenic factor PEA3 regulates OCT4 gene expression as a transcriptional activator.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA005768
Anti-ETV4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32263
Anti-ETV4 antibody produced in rabbit, affinity isolated antibody
SAB4500817
Anti-ETV4 antibody produced in rabbit, affinity isolated antibody
AV44026
Anti-SLC22A3 antibody produced in rabbit, affinity isolated antibody
HPA029750
Anti-SLC22A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502815
Anti-SLC22A3 antibody produced in rabbit, affinity isolated antibody
SAB2106660
Anti-SLC22A3 antibody produced in rabbit, affinity isolated antibody
SAB1406815
Anti-SLC22A8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA044174
Anti-SLC22A8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0002118M1
Monoclonal Anti-ETV4 antibody produced in mouse, clone 3G9-1B9, purified immunoglobulin, buffered aqueous solution
SAB1403795
Monoclonal Anti-ETV4 antibody produced in mouse, clone 1A3-1D3, purified immunoglobulin, buffered aqueous solution
SAB5300472
Monoclonal Anti-ETV4 antibody produced in mouse, clone 1A2G3, ascites fluid
SAB1403021
Monoclonal Anti-SLC22A8 antibody produced in mouse, clone 3C11, purified immunoglobulin, buffered aqueous solution