EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Ascochlorin derivatives from the leafhopper pathogenic fungus Microcera sp. BCC 17074.
PMID 24984794
Abstract
Two new ascochlorin derivatives, nectchlorins A (1) and B (2), together with eight known compounds (3-10), were isolated from cultures of the leafhopper pathogen Microcera sp. BCC 17074. The structures were elucidated on the basis of NMR spectroscopic and mass spectrometry data. The absolute configuration of 2 was determined by application of the modified Mosher's method. The absolute configuration of LL-Z 1272α epoxide (9), which is a plausible biosynthetic precursor of ascochlorins, was established by chemical correlations. Cytotoxic activities of these ascochlorin derivatives were evaluated.