EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Antagonizing c-Cbl enhances EGFR-dependent corneal epithelial homeostasis.
PMID 24985478
Abstract
In many cell types, the E3 ubiquitin ligase, c-Cbl, induces ligand-dependent ubiquitylation of the epidermal growth factor receptor (EGFR) and targets the receptor for lysosomal degradation. The goal of this study was to determine whether c-Cbl is a negative regulator of EGFR in the corneal epithelium and if it can be inhibited to promote corneal epithelial homeostasis. Expression and activity of c-Cbl were blocked in immortalized human corneal epithelial cells (hTCEpi) using RNAi and pharmacological agents ([4-amino-5-(4-methylphenyl)-7-(t-butyl)pyrazolo-d-3,4-pyrimidine] or PP1). Following c-Cbl inhibition, cells were assessed for ligand-dependent receptor ubiquitylation, receptor phosphorylation, and in vitro wound healing. Subsequent experiments used PP1 in hTCEpi cells and monitored in vivo murine corneal epithelial wound healing. Knockdown and inhibition of c-Cbl decreased ligand-dependent ubiquitylation of the EGFR and prolonged receptor activity as measured by tyrosine phosphorylation. Further, these treatments also increased the extent of ligand-dependent corneal epithelial wound healing in vitro and in vivo. Manipulating the duration of EGFR activity can enhance the rate of restoration of the corneal epithelial layer. Based on our findings, c-Cbl is a new therapeutic target to enhance EGFR-mediated corneal epithelial homeostasis that bypasses the limitations of previous approaches.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50553
α-tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Ca3O8P2
53737
α-tri-Calcium phosphate, puriss. p.a., ≥75% α-phase basis (sinterted powder)
Ca3O8P2
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300480
Anti-EGFR (Ab-1197) antibody produced in rabbit, affinity isolated antibody
SAB4300453
Anti-EGFR (Ab-678) antibody produced in rabbit, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
SAB4300481
Anti-EGFR (Ab-869) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018530
Anti-EGFR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500720
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
13204
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Ca3O8P2
49963
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Ca3O8P2
21218
Calcium phosphate, purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)
Ca3O8P2
WH0001956M2 Monoclonal Anti-EGFR antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
AMAB90816
Monoclonal Anti-EGFR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0815, purified immunoglobulin, buffered aqueous glycerol solution
E2760
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
E3138
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
E0532 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
P9405
O-Phospho-L-tyrosine
C9H12NO6P
PLA0101
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.