EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of perfluorinated alkyl acids in corn, popcorn and popcorn bags before and after cooking by focused ultrasound solid-liquid extraction, liquid chromatography and quadrupole-time of flight mass spectrometry.
PMID 24986069
Abstract
An analytical method is proposed to determine ten perfluorinated alkyl acids (PFAAs) [nine perfluorocarboxylic acids (PFCAs) and perfluorooctane sulfonate (PFOS)] in corn, popcorn and microwave popcorn packaging by focused ultrasound solid-liquid extraction (FUSLE) and ultra high performance liquid chromatography (UHPLC) coupled to quadrupole-time of flight mass spectrometry (QTOF-MS/MS). Selected PFAAs were extracted efficiently in only one 10-s cycle by FUSLE, a simple, safe and inexpensive technique. The developed method was validated for microwave popcorn bags matrix as well as corn and popcorn matrices in terms of linearity, matrix effect error, detection and quantification limits, repeatability and recovery values. The method showed good accuracy with recovery values around 100% except for the lowest chain length PFAAs, satisfactory reproducibility with RSDs under 16%, and sensitivity with limits of detection in the order of hundreds picograms per gram of sample (between 0.2 and 0.7ng/g). This method was also applied to the analysis of six microwave popcorn bags and the popcorn inside before and after cooking. PFCAs contents between 3.50ng/g and 750ng/g were found in bags, being PFHxA (perfluorohexanoic acid) the most abundant of them. However, no PFAAs were detected either corn or popcorn, therefore no migration was assumed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
77283
Heptadecafluorooctanesulfonic acid solution, ~40% in H2O (T)
C8HF17O3S
33607
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
C8HF17O3S
77288
Heptadecafluorooctanesulfonic acid tetraethylammonium salt, purum, ≥98.0% (T)
C16H20F17NO3S
365289
Heptadecafluorooctanesulfonic acid tetraethylammonium salt, 98%
C16H20F17NO3S
77249
Heptafluorobutyric acid, ≥99.0% (GC)
C4HF7O2
52411
Heptafluorobutyric acid, for ion chromatography, ≥99.5% (GC)
C4HF7O2
164194
Heptafluorobutyric acid, 98%
C4HF7O2
49540
Heptafluorobutyric acid solution, 0.5 M in H2O, for ion pair chromatography
C4HF7O2
33824
Pentadecafluorooctanoic acid, analytical standard
C8HF15O2
33603
Pentadecafluorooctanoic acid solution, 100 μg/mL in methanol, analytical standard
C8HF15O2
177741
Perfluorodecanoic acid, 98%
C10HF19O2
342041
Perfluoroheptanoic acid, 99%
C7HF13O2
394459
Perfluorononanoic acid, 97%
C9HF17O2
171468
Perfluorooctanoic acid, 96%
C8HF15O2
396575
Perfluoropentanoic acid, 97%
C5HF9O2
446777
Perfluoroundecanoic acid, 95%
C11HF21O2
71539
Sodium formate, BioUltra, ≥99.0% (NT)
CHNaO2
67253
Sodium formate, analytical standard
CHNaO2
456020
Sodium formate, 99.998% trace metals basis
CHNaO2
587036
Sodium formate-12C, 99.9 atom % 12C
12CHNaO2
406449
Tricosafluorododecanoic acid, 95%
C12HF23O2
29226
Undecafluorohexanoic acid, ≥97.0% (T)
C6HF11O2