PloS one
Expression of the genetic suppressor element 24.2 (GSE24.2) decreases DNA damage and oxidative stress in X-linked dyskeratosis congenita cells.
PMID 24987982
Abstract
The predominant X-linked form of Dyskeratosis congenita results from mutations in DKC1, which encodes dyskerin, a protein required for ribosomal RNA modification that is also a component of the telomerase complex. We have previously found that expression of an internal fragment of dyskerin (GSE24.2) rescues telomerase activity in X-linked dyskeratosis congenita (X-DC) patient cells. Here we have found that an increased basal and induced DNA damage response occurred in X-DC cells in comparison with normal cells. DNA damage that is also localized in telomeres results in increased heterochromatin formation and senescence. Expression of a cDNA coding for GSE24.2 rescues both global and telomeric DNA damage. Furthermore, transfection of bacterial purified or a chemically synthesized GSE24.2 peptide is able to rescue basal DNA damage in X-DC cells. We have also observed an increase in oxidative stress in X-DC cells and expression of GSE24.2 was able to diminish it. Altogether our data indicated that supplying GSE24.2, either from a cDNA vector or as a peptide reduces the pathogenic effects of Dkc1 mutations and suggests a novel therapeutic approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB3560
Anti-8-Oxoguanine Antibody, clone 483.15, 100 µL, Anti-8-Oxoguanine Antibody, clone 483.15 detects level of 8-Oxoguanine & has been published & validated for use in ELISA & IC.
05-636
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, 200 µg, Anti-phospho-Histone H2A.X (Ser139), clone JBW301 is a well published Mouse Monoclonal Antibody validated in ChIP, ICC, IF, WB. This purified mAb is highly specific for phospho-Histone H2A.X (Ser139) also known as H2AXS139p.
GW21178 Anti-ATM antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500147
Anti-ATM antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501968
Anti-ATM antibody produced in rabbit, affinity isolated antibody
A6218 Anti-ATM antibody, Mouse monoclonal, clone SYML6A10, purified from hybridoma cell culture
A6093 Anti-ATM antibody, Mouse monoclonal, clone SYR6D4, purified from hybridoma cell culture
D7008
Dihydroethidium, ≥95%
C21H21N3
37291
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
C21H21N3
PHR1359
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
C10H17N3O6S
Y0000517
Glutathione, European Pharmacopoeia (EP) Reference Standard
C10H17N3O6S
PLA0045
Goat anti-ATM Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB1402696
Monoclonal Anti-ATM antibody produced in mouse, clone 2C10, purified immunoglobulin, buffered aqueous solution
A1106
Monoclonal Anti-ATM antibody produced in mouse, ~2 mg/mL, clone MAT3-4G10/8, purified immunoglobulin, PBS solution
PLA0086
Rabbit anti-ATM Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.