PloS one
Mammary gland specific knockdown of the physiological surge in Cx26 during lactation retains normal mammary gland development and function.
PMID 24988191
Abstract
Connexin26 (Cx26) is the major Cx protein expressed in the human mammary gland and is up-regulated during pregnancy while remaining elevated throughout lactation. It is currently unknown if patients with loss-of-function Cx26 mutations that result in hearing loss and skin diseases have a greater susceptibility to impaired breast development. To investigate if Cx26 plays a critical role in mammary gland development and differentiation, a novel Cx26 conditional knockout mouse model was generated by crossing Cx26fl/fl mice with mice expressing Cre under the β-Lactoglobulin promoter. Conditional knockdown of Cx26 from the mammary gland resulted in a dramatic reduction in detectable gap junction plaques confirmed by a significant ∼65-70% reduction in Cx26 mRNA and protein throughout parturition and lactation. Interestingly, this reduction was accompanied by a decrease in mammary gland Cx30 gap junction plaques at parturition, while no change was observed for Cx32 or Cx43. Whole mount, histological and immunofluorescent assessment of breast tissue revealed comparatively normal lobuloalveolar development following pregnancy in the conditionally knockdown mice compared to control mice. In addition, glands from genetically-modified mice were capable of producing milk proteins that were evident in the lumen of alveoli and ducts at similar levels as controls, suggesting normal gland function. Together, our results suggest that low levels of Cx26 expression throughout pregnancy and lactation, and not the physiological surge in Cx26, is sufficient for normal gland development and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB374
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, 200 µg, Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5 is a well published and extensively characterized monoclonal antibody. This purified mAb detects Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) & has been published & validated for use in ELISA, IP, IC, IF, IH & WB.
SAB4501173
Anti-Connexin 43 antibody produced in rabbit, affinity isolated antibody
C3595 Anti-Connexin-32 (106-124) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
C3470
Anti-Connexin-32 (265-279) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300504
Anti-GJA1 (Ab-367) antibody produced in rabbit, affinity isolated antibody
AV36598
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody
SAB2900237
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100918
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody
SAB1300557 Anti-GJA3 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2100919
Anti-GJA3 antibody produced in rabbit, affinity isolated antibody
SAB4501178
Anti-GJA3 antibody produced in rabbit, affinity isolated antibody
HPA010663
Anti-GJB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV36600
Anti-GJB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2104320
Anti-GJB1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV36608
Anti-GJB2 antibody produced in rabbit, IgG fraction of antiserum
SAB2104321
Anti-GJB6 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1307074
ANTI-GJB6(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1402787
Monoclonal Anti-GJB2 antibody produced in mouse, clone 1C6, purified immunoglobulin, buffered aqueous solution