EMAIL THIS PAGE TO A FRIEND
Scientific reports
Growth arrest and forced differentiation of human primary glioblastoma multiforme by a novel small molecule.
PMID 24989033
Abstract
Glioblastoma multiforme is the most common malignant brain tumor in adults, with an average survival of less than one year due to its resistance to therapy. Recent studies reported that GBM initiates from CD133-expressing cancer stem cells (CSC). However, the efficacy of CSC targeting is limited. A newly developed approach in cancer treatment is the forced differentiation of cancer cells. Here, we show that the treatment of the novel small molecule, CG500354, into CD133-expressing human primary GBM cells induces growth arrest by cell cycle regulators, p53, p21, p27 and phase-specific cyclins, and neural differentiation, as confirmed by neural progenitor/precursor markers, nestin, GFAP and Tuj1. When GBM-derived cells caused the tumors in NOD/SCID mice, CG500354 induced GBM-derived cells differentiation into Tuj1 and GFAP expressing cells. We next demonstrated that CG500354 plays a tumor-suppressive role via cAMP/CREB signaling pathway. CG500354 increases not only the extracellular cAMP level but also the protein level of PKA and CREB. Additionally, both mimetic substances, Forskolin and Rolipram, revealed comparable results with CG500354. Our findings indicate that induction of growth arrest and neural differentiation via cAMP/CREB signaling pathway by CG500354 treatment suggests the novel targeting of PDE4D in the development of new drugs for brain tumor therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB374
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, 200 µg, Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5 is a well published and extensively characterized monoclonal antibody. This purified mAb detects Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) & has been published & validated for use in ELISA, IP, IC, IF, IH & WB.
MAB5326
Anti-Nestin Antibody, clone 10C2, 100 µg, This Anti-Nestin Antibody, clone 10C2 is validated for use in IC, IH, IH(P), WB for the detection of Nestin.
SAB4503908
Anti-phospho-CREB (pSer121) antibody produced in rabbit, affinity isolated antibody
C9102 Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4503757
Anti-phospho-CREB (pSer142) antibody produced in rabbit, affinity isolated antibody
SAB4504577
Anti-phospho-CREB (pThr100) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300217
Anti-phospho-CREB1 (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB2107606
Anti-PROM1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
R6520
Rolipram, solid, ≥98% (HPLC)
C16H21NO3