EMAIL THIS PAGE TO A FRIEND
Human fertility (Cambridge, England)
Oral antioxidants supplementation for women with unexplained infertility undergoing ICSI/IVF: randomized controlled trial.
PMID 24989721
Abstract
Good oocyte quality and maturity are important prerequisites for high fertilization and implantation rates in IVF/ICSI treatment cycles. Reactive oxygen species (ROS) are produced within ovarian follicles, especially during the ovulation process, and increased ROS activity may be a cause of impaired oocyte maturation and higher rate of failure of IVF/ICSI cycles. RCT evaluating the effect of antioxidant supplementation on ICSI/IVF outcomes. Two hundred and eighteen women with unexplained subfertility undergoing IVF/ICSI were randomized into two groups. The study group (n = 112) received daily oral antioxidants in the form of multivitamins and minerals (amino acid chelated) while the control group (n = 106) did not. Main outcomes were number of mature metaphase II (MII) oocytes and clinical pregnancy rate. There were no significant changes between the groups as regards age, BMI, basal FSH, number of mature (MII) oocytes (12.7 ± 9.4 vs. 13.2 ± 8.6, P = 0.7) and clinical pregnancy rate per woman randomized (38% vs. 34%; [OR = 1.2; 95% CI, 0.70-2.11]. Oral antioxidants in the form of a combination of multivitamins and minerals (amino acid chelated) did not improve oocyte quality and pregnancy rates in women with unexplained infertility undergoing IVF/ICSI treatment.