EMAIL THIS PAGE TO A FRIEND
Lab on a chip
A lab-in-a-briefcase for rapid prostate specific antigen (PSA) screening from whole blood.
PMID 24989886
Abstract
We present a new concept for rapid and fully portable prostate specific antigen (PSA) measurements, termed "lab-in-a-briefcase", which integrates an affordable microfluidic ELISA platform utilising a melt-extruded fluoropolymer microcapillary film (MCF) containing an array of 10 200 μm internal diameter capillaries, a disposable multi-syringe aspirator (MSA), a sample tray pre-loaded with all of the required immunoassay reagents, and a portable film scanner for colorimetric signal digital quantification. Each MSA can perform 10 replicate microfluidic immunoassays on 8 samples, allowing 80 measurements to be made in less than 15 minutes based on semi-automated operation, without the need of additional fluid handling equipment. The assay was optimised for the measurement of a clinically relevant range of PSA of 0.9 to 60.0 ng ml(-1) in 15 minutes with CVs on the order of 5% based on intra-assay variability when read using a consumer flatbed film scanner. The PSA assay performance in the MSA remained robust in undiluted or 1 : 2 diluted human serum or whole blood, and the matrix effect could simply be overcome by extending sample incubation times. The PSA "lab-in-a-briefcase" is particularly suited to a low-resource health setting, where diagnostic labs and automated immunoassay systems are not accessible, by allowing PSA measurement outside the laboratory using affordable equipment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0729 Human KLK3 / Prostate-specific Antigen ELISA Kit
RAB0331
Human PSA-total ELISA Kit, for serum, plasma, cell culture supernatant and urine
95314
Hydrogen peroxide–Urea adduct, purum p.a., "rapid-soluble", tablet (1 g each)
CH4N2O · H2O2
U8879
Hydrogen peroxide–Urea adduct, tablet
CH4N2O · H2O2
04078
Hydrogen peroxide–Urea adduct, powder, 15-17% active oxygen basis
CH4N2O · H2O2
P6662
o-Phenylenediamine dihydrochloride, tablet, 1 mg substrate per tablet
C6H8N2 · 2HCl
P6787
o-Phenylenediamine dihydrochloride, tablet, 2 mg substrate per tablet
C6H8N2 · 2HCl
P8806
o-Phenylenediamine dihydrochloride, tablet, 3 mg substrate per tablet
C6H8N2 · 2HCl
P8787
o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet
C6H8N2 · 2HCl
P3804
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
C6H8N2 · 2HCl
P6912
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
C6H8N2 · 2HCl
P8287
o-Phenylenediamine dihydrochloride, tablet, 10 mg substrate per tablet
C6H8N2 · 2HCl
P4664
o-Phenylenediamine dihydrochloride, tablet, 15 mg substrate per tablet
C6H8N2 · 2HCl
P7288
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
C6H8N2 · 2HCl
P8412
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet
C6H8N2 · 2HCl
P1063
o-Phenylenediamine dihydrochloride, tablet, 60 mg substrate per tablet
C6H8N2 · 2HCl
289132
Urea hydrogen peroxide, 97%
CH4N2O · H2O2