EMAIL THIS PAGE TO A FRIEND
Retrovirology
Human immature Langerhans cells restrict CXCR4-using HIV-1 transmission.
PMID 24990163
Abstract
Sexual transmission is the main route of HIV-1 infection and the CCR5-using (R5) HIV-1 is predominantly transmitted, even though CXCR4-using (X4) HIV-1 is often abundant in chronic HIV-1 patients. The mechanisms underlying this tropism selection are unclear. Mucosal Langerhans cells (LCs) are the first immune cells to encounter HIV-1 and here we investigated the role of LCs in selection of R5 HIV-1 using an ex vivo epidermal and vaginal transmission models. Immature LCs were productively infected by X4 as well as R5 HIV-1. However, only R5 but not X4 viruses were selectively transmitted by immature LCs to T cells. Transmission of HIV-1 was depended on de novo production of HIV-1 in LCs, since it could be inhibited by CCR5 fusion inhibitors as well as reverse transcription inhibitors. Notably, the activation state of LCs affected the restriction in X4 HIV-1 transmission; immune activation by TNF facilitated transmission of X4 as well as R5 HIV-1. These data suggest that LCs play a crucial role in R5 selection and that immature LCs effectively restrict X4 at the level of transmission.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405633
Anti-CCR5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
PRS1112
Anti-CCR5 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2100370
Anti-CCR5 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB1407414
Anti-CD207 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA011216
Anti-CD207 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401669
Anti-CD207 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1401467
Anti-CD83 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411641
Anti-CD83 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1411654
Anti-CD86 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21075 Anti-CXCR4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C3116
Anti-CXCR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500383
Anti-CXCR4 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
C5973 Monoclonal Anti-CCR-5 antibody produced in mouse, clone 45523, purified immunoglobulin, lyophilized powder
WH0009308M1
Monoclonal Anti-CD83 antibody produced in mouse, clone 3G10-1F4, purified immunoglobulin, buffered aqueous solution
SAB1404701
Monoclonal Anti-CD83 antibody produced in mouse, clone 3F3, purified immunoglobulin, buffered aqueous solution
WH0000942M1 Monoclonal Anti-CD86 antibody produced in mouse, clone 3D7, purified immunoglobulin, buffered aqueous solution
SAB4700144 Monoclonal Anti-CD86 antibody produced in mouse, clone BU63, purified immunoglobulin, buffered aqueous solution
SAB4700560 Monoclonal Anti-Cd86 antibody produced in rat, clone GL-1, purified immunoglobulin, buffered aqueous solution
C6598 Monoclonal Anti-CXCR4 (Fusin) antibody produced in mouse, clone 44716, purified immunoglobulin, lyophilized powder
WH0007852M2
Monoclonal Anti-CXCR4 antibody produced in mouse, clone 1F8, ascites fluid, buffered aqueous solution
SAB1404560
Monoclonal Anti-CXCR4, (N-terminal) antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution