EMAIL THIS PAGE TO A FRIEND
Immunology
Inhibition of CD1d-mediated antigen presentation by the transforming growth factor-β/Smad signalling pathway.
PMID 24990409
Abstract
CD1d-mediated lipid antigen presentation activates a subset of innate immune lymphocytes called invariant natural killer T (NKT) cells that, by virtue of their potent cytokine production, bridge the innate and adaptive immune systems. Transforming growth factor (TGF-β) is a known immune modulator that can activate the mitogen-activated protein kinase p38; we have previously shown that p38 is a negative regulator of CD1d-mediated antigen presentation. Several studies implicate a role for TGF-β in the activation of p38. Therefore, we hypothesized that TGF-β would impair antigen presentation by CD1d. Indeed, a dose-dependent decrease in CD1d-mediated antigen presentation and impairment of lipid antigen processing was observed in response to TGF-β treatment. However, it was found that this inhibition was not through p38 activation. Instead, Smads 2, 3 and 4, downstream elements of the TGF-β canonical signalling pathway, contributed to the observed effects. In marked contrast to that observed with CD1d, TGF-β was found to enhance MHC class II-mediated antigen presentation. Overall, these results suggest that the canonical TGF-β/Smad pathway negatively regulates an important arm of the host's innate immune responsesxa0-xa0CD1d-mediated lipid antigen presentation to NKT cells.