EMAIL THIS PAGE TO A FRIEND
International journal of radiation biology
Dose-effect of ionizing radiation-induced PIG3 gene expression alteration in human lymphoblastoid AHH-1 cells and human peripheral blood lymphocytes.
PMID 24991881
Abstract
To identify new ionizing radiation (IR)-sensitive genes and observe the dose-effect of gene expression alteration (GEA) induced by IR. Microarray was used to screen the differentially expressed genes in human lymphoblastoid cells (AHH-1) using three doses of (60)Co γ-rays (0.5-8 Gy at 1 Gy/min). Given that p53-inducible gene 3 (PIG3) was consistently upregulated, the GEA of PIG3 in AHH-1 cells and human peripheral blood lymphocytes (HPBL) induced by γ-rays (1 Gy/min) was measured at messenger RNA (mRNA) and protein levels. The GEA of PIG3 in AHH-1 cells exposed to neutron radiation (californium-252, 0.073 Gy/min) was also quantified. PIG3 was one of the seven differentially expressed genes found in the microarray analysis. The PIG3 mRNA and protein levels in AHH-1 cells were significantly increased from 1-10 Gy of γ-rays 8-72 h or 8-168 h after exposure, respectively. The enhancement was also observed in AHH-1 cells from 0.4-1.6 Gy of neutrons 48 h post-irradiation. The PIG3 mRNA levels (mRNA copy numbers) in HPBL were significantly increased from 1-8 Gy of γ-rays within 4-24 h post-irradiation, but the highest increase in signal-to-noise responsiveness is approximately two-fold, which was less than that of AHH-1 (approximately 20-fold). IR can upregulate the PIG3 gene expression in AHH-1 and HPBL in the early phase after exposure; however, the IR induced expression levels of PIG3 are greater in AHH-1 than HPBL.