EMAIL THIS PAGE TO A FRIEND
British journal of cancer
The role of TGF-β1-miR-21-ROS pathway in bystander responses induced by irradiated non-small-cell lung cancer cells.
PMID 24992582
Abstract
Many studies have indicated an important implication of radiation-induced bystander effects (RIBEs) in cancer radiotherapy, but the detailed signalling remains unclear. The roles of tumour growth factor-beta1 (TGF-β1) and miR-21 in medium-mediated RIBEs in H1299 non-small-cell lung cancer cells were investigated using DNA damage, changes in proliferation and levels of reactive oxygen species (ROS) as end points. SB431542, a specific inhibitor of TGF-β type 1 receptor kinases, was used to inhibit TGF-β1 pathways in irradiated and bystander cells. Exogenous miR-21 regulation was achieved through inhibitor or mimic transfection. Compared with relative sham-radiation-conditioned medium, radiation-conditioned medium (RCM) from irradiated cells 1 h post radiation (1-h RCM) caused an increase in ROS levels and DNA damage in bystander cells, while 18-h RCM induced cell cycle delay and proliferation inhibition. All these effects were eliminated by TGF-βR1 inhibition. One-hour RCM upregulated miR-21 expression in bystander cells, and miR-21 inhibitor abolished bystander oxidative stress and DNA damage. Eighteen-hour RCM downregulated miR-21 of bystander cells, and miR-21 mimic eliminated bystander proliferation inhibition. Furthermore, the dysregulation of miR-21 was attenuated by TGF-βR1 inhibition. The TGF-β1-miR-21-ROS pathway of bystander cells has an important mediating role in RIBEs in H1299 cells.