EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Hepatic, intestinal, renal, and plasma hydrolysis of prodrugs in human, cynomolgus monkey, dog, and rat: implications for in vitro-in vivo extrapolation of clearance of prodrugs.
PMID 24994071
Abstract
Hydrolysis plays an important role in metabolic activation of prodrugs. In the current study, species and in vitro system differences in hepatic and extrahepatic hydrolysis were investigated for 11 prodrugs. Ten prodrugs in the data set are predominantly hydrolyzed by carboxylesterases (CES), whereas olmesartan medoxomil is also metabolized by carboxymethylenebutenolidase (CMBL) and paraoxonase. Metabolic stabilities were assessed in cryopreserved hepatocytes, liver S9 (LS9), intestinal S9 (IS9), kidney S9 (KS9), and plasma from human, monkey, dog, and rat. Of all the preclinical species investigated, monkey intrinsic hydrolysis clearance obtained in hepatocytes (CLint,hepatocytes) were the most comparable to human hepatocyte data. Perindopril and candesartan cilexetil showed the lowest and highest CLint,hepatocytes, respectively, regardless of the species investigated. Scaled intrinsic hydrolysis clearance obtained in LS9 were generally higher than CLint,hepatocytes in all species investigated, with the exception of dog. In the case of human and dog intestinal S9, hydrolysis intrinsic clearance could not be obtained for CES1 substrates, but hydrolysis for CES2 and CMBL substrates was detected in IS9 and KS9 from all species. Pronounced species differences were observed in plasma; hydrolysis of CES substrates was only evident in rat. Predictability of human hepatic intrinsic clearance (CLint,h) was assessed for eight CES1 substrates using hepatocytes and LS9; extrahepatic hydrolysis was not considered due to high stability of these prodrugs in intestinal and kidney S9. On average, predicted oral CLint,h from hepatocyte data represented 20% of the observed value; the underprediction was pronounced for high-clearance prodrugs, consistent with the predictability of cytochrome P450/conjugation clearance from this system. Prediction bias was less apparent with LS9, in particular for high-clearance prodrugs, highlighting the application of this in vitro system for investigation of prodrugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001038
Benazepril for system suitability, European Pharmacopoeia (EP) Reference Standard
C24H28N2O5 · HCl
B0935
Benazepril hydrochloride, ≥98% (HPLC), solid
C24H28N2O5 · HCl
Y0001025
Benazepril hydrochloride, European Pharmacopoeia (EP) Reference Standard
C24H28N2O5 · HCl
SML0245
Candesartan cilexetil, ≥98% (HPLC)
C33H34N6O6
Y0001388
Candesartan cilexetil, European Pharmacopoeia (EP) Reference Standard
C33H34N6O6
Y0001387
Candesartan cilexetil for peak identification, European Pharmacopoeia (EP) Reference Standard
C33H34N6O6
Y0001389
Candesartan cilexetil for system suitability, European Pharmacopoeia (EP) Reference Standard
C33H34N6O6
SML0284
Mycophenolate mofetil, ≥98% (HPLC)
C23H31NO7
1448956
Mycophenolate mofetil, United States Pharmacopeia (USP) Reference Standard
C23H31NO7
Y0000489
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard
C23H31NO7
PHR1451
Mycophenolate Mofetil, Pharmaceutical Secondary Standard; Certified Reference Material
C23H31NO7
Y0000519
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard
C23H31NO7
1479304
Oseltamivir phosphate, United States Pharmacopeia (USP) Reference Standard
C16H28N2O4 ·H3PO4
Y0001337
Oseltamivir phosphate (impurity B free), European Pharmacopoeia (EP) Reference Standard
C16H28N2O4 ·H3PO4
P0094
Perindopril erbumine
C19H32N2O5 · C4H11N
Y0000771
Perindopril for peak identification, European Pharmacopoeia (EP) Reference Standard
C19H32N2O5 · C4H11N
Y0000207
Perindopril for stereochemical purity, European Pharmacopoeia (EP) Reference Standard
C19H32N2O5 · C4H11N
Y0000238
Perindopril tert-butylamine, European Pharmacopoeia (EP) Reference Standard
C19H32N2O5 · C4H11N
Y0001502
Quinapril for peak identification, European Pharmacopoeia (EP) Reference Standard
C25H30N2O5 · HCl
Y0001503
Quinapril for system suitability, European Pharmacopoeia (EP) Reference Standard
C25H30N2O5 · HCl
Q0632
Quinapril hydrochloride, ≥98% (HPLC), solid
C25H30N2O5 · HCl
Y0001491
Quinapril hydrochloride, European Pharmacopoeia (EP) Reference Standard
C25H30N2O5 · HCl
R0404
Ramipril, ≥98% (HPLC)
C23H32N2O5
1598303
Ramipril, United States Pharmacopeia (USP) Reference Standard
C23H32N2O5
R0145000
Ramipril, European Pharmacopoeia (EP) Reference Standard
C23H32N2O5
PHR1446
Ramipril, Pharmaceutical Secondary Standard; Certified Reference Material
C23H32N2O5