EMAIL THIS PAGE TO A FRIEND
British journal of haematology
Wnt inhibition leads to improved chemosensitivity in paediatric acute lymphoblastic leukaemia.
PMID 24995804
Abstract
While childhood acute lymphoblastic leukaemia (ALL) is now highly curable, the dismal prognosis for children who relapse warrants novel therapeutic approaches. Previously, using an integrated genomic analysis of matched diagnosis-relapse paired samples, we identified overactivation of the Wnt pathway as a possible mechanism of recurrence. To validate these findings and document whether Wnt inhibition may sensitize cells to chemotherapy, we analysed the expression of activated β-catenin (and its downstream target BIRC5) using multiparameter phosphoflow cytometry and tested the efficacy of a recently developed Wnt inhibitor, iCRT14, in ALL cell lines and patient samples. We observed increased activation of β-catenin at relapse in 6/10 patients. Furthermore, treatment of leukaemic cell lines with iCRT14 led to significant downregulation of Wnt target genes and combination with traditional chemotherapeutic drugs resulted in a synergistic decrease in viability as well as a significant increase in apoptotic cell death. Finally, pre-treatment of purified blasts from patients with relapsed leukaemia with the Wnt inhibitor followed by exposure to prednisolone, restored chemosensitivity in these cells. Our results demonstrate that overactivation of the Wnt pathway may contribute to chemoresistance in relapsed childhood ALL and that Wnt-inhibition may be a promising therapeutic approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0203
iCRT14, ≥98% (HPLC)
C21H17N3O2S