EMAIL THIS PAGE TO A FRIEND
Regulatory toxicology and pharmacology : RTP
Flatworm models in pharmacological research: the importance of compound stability testing.
PMID 24999090
Abstract
Flatworms possess adult pluripotent stem cells, which make them extraordinary experimental model organisms to assess in vivo the undesirable effects of substances on stem cells. Currently, quality practices, implying evaluation of the stability of the test compound under the proposed experimental conditions, are uncommon in this research field. Nevertheless, performing a stability study during the rational design of in vivo assay protocols will result in more reliable assay results. To illustrate the influence of the stability of the test substance on the final experimental outcome, we performed a short-term International Conference on Harmonization (ICH)-based stability study of cyclophosphamide in the culture medium, to which a marine flatworm model Macrostomum lignano is exposed. Using a validated U(H)PLC method, it was demonstrated that the cyclophosphamide concentration in the culture medium at 20°C is lowered to 80% of the initial concentration after 21days. The multiwell plates, flatworms and diatoms, as well as light exposure, did not influence significantly the cyclophosphamide concentration in the medium. The results of the stability study have practical implications on the experimental set-up of the carcinogenicity assay like the frequency of medium renewal. This case study demonstrates the benefits of applying appropriate quality guidelines already during fundamental research increasing the credibility of the results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0768
Cyclophosphamide monohydrate, bulk package
C7H15Cl2N2O2P · H2O
C7397
Cyclophosphamide monohydrate, ISOPAC®
C7H15Cl2N2O2P · H2O
93813
Cyclophosphamide monohydrate, analytical standard
C7H15Cl2N2O2P · H2O