EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Preparation and application of sol-gel acrylate and methacrylate solid-phase microextraction fibres for gas chromatographic analysis of organoarsenic compounds.
PMID 25000858
Abstract
Novel solid-phase microextraction (SPME) fibres containing methyl, ethyl, butyl acrylate and methacrylate were first prepared by a sol-gel technique and investigated for determination of selected organoarsenic compounds (lewisite, methyldichloroarsine, phenyldichloroarsine, diphenylchloroarsine and triphenylarsine) from water samples. The influence of sorption and desorption temperature and time for extraction efficiency were examined. The best new fibre coatings (methyl acrylate (MA), methyl methacrylate (MMA) and combination of methyl acrylate and methacrylate (MA/MMA)) for analysis of organoarsenic compounds were selected and compared with commercial fibres. The distribution coefficients Kfs were determined for the best novel fibres and for absorption commercial fibres. The highest Kfs value were obtained for MA/MMA and MMA fibres and were respectively 9458 and 6561 for lewisite and 6458 and 5884 for triphenylarsine. The limit of detection and quantification were determined for the three laboratory obtained fibres (MA, MMA and MA/MMA). LODs for tested fibres, at a signal-to-noise of 3, were 0.03-0.3 ng mL(-1). LOQs for selected coatings, at signal-to-noise of 10, were 0.1-0.8 ng mL(-1). The relative standard deviations (RSD) for all measurements were 4.3-6.5% (n=9) and relative errors were 2.5-5%. The laboratory obtained fibres were used for environmental analysis of pore water samples from the Baltic Sea.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
95446
1-Octanol, analytical standard
C8H18O
74850
1-Octanol, puriss., ≥99.5% (GC)
C8H18O
W280003
1-Octanol, ≥98%, FCC, FG
C8H18O
W280011
1-Octanol, natural, ≥98%, FCC
C8H18O
06958
Butyl acrylate, analytical standard
C7H12O2
234923
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor
C7H12O2
235865
Butyl methacrylate, 99%, contains 10 ppm monomethyl ether hydroquinone as inhibitor
C8H14O2
Y0001487
Butyl methacrylate, European Pharmacopoeia (EP) Reference Standard
C8H14O2
W221503
Butyl sulfide, ≥95%, FG
C8H18S
51491
Dibutyl sulfide, ≥98.0% (GC)
C8H18S
B101796
Dibutyl sulfide, 96%
C8H18S
76130
Ethyl acrylate, analytical standard
C5H8O2
E9706
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
C5H8O2
W241806
Ethyl acrylate, ≥99.5%, stabilized, FG
C5H8O2
234893
Ethyl methacrylate, contains 15-20 ppm monomethyl ether hydroquinone as inhibitor, 99%
C6H10O2
4S42585
Ethyl methacrylate, analytical standard
C6H10O2
2854
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
C10H14N2Na2O8
E7889
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
C10H14N2Na2O8
I26809
Isopropyl sulfide, 99%
C6H14S
76778
Methyl acrylate, analytical standard
C4H6O2
M27301
Methyl acrylate, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor
C4H6O2
Y0001488
Methyl methacrylate, European Pharmacopoeia (EP) Reference Standard
C5H8O2
110175
Tetraethylene glycol, 99%
C8H18O5
T81906
Triphenylarsine, 97%
C18H15As