EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
SIRT1 inhibition during the hypoinflammatory phenotype of sepsis enhances immunity and improves outcome.
PMID 25001863
Abstract
Mechanism-based sepsis treatments are unavailable, and their incidence is rising worldwide. Deaths occur during the early acute phase of hyperinflammation or subsequent postacute hypoinflammatory phase with sustained organ failure. The acute sepsis phase shifts rapidly, and multiple attempts to treat early excessive inflammation have uniformly failed. We reported in a sepsis cell model and human sepsis blood leukocytes that nuclear NAD+ sensor SIRT1 deacetylase remodels chromatin at specific gene sets to switch the acute-phase proinflammatory response to hypoinflammatory. Importantly, SIRT1 chromatin reprogramming is reversible, suggesting that inhibition of SIRT1 might reverse postacute-phase hypoinflammation. We tested this concept in septic mice, using the highly specific SIRT1 inhibitor EX-527, a small molecule that closes the NAD+ binding site of SIRT1. Strikingly, when administered 24 h after sepsis, all treated animals survived, whereas only 40% of untreated mice survived. EX-527 treatment reversed the inability of leukocytes to adhere at the small intestine MVI, reversed in vivo endotoxin tolerance, increased leukocyte accumulation in peritoneum, and improved peritoneal bacterial clearance. Mechanistically, the SIRT1 inhibitor restored repressed endothelial E-selectin and ICAM-1 expression and PSGL-1 expression on the neutrophils. Systemic benefits of EX-527 treatment included stabilized blood pressure, improved microvascular blood flow, and a shift toward proimmune macrophages in spleen and bone marrow. Our findings reveal that modifying the SIRT1 NAD+ axis may provide a novel way to treat sepsis in its hypoinflammatory phase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-131
Anti-Sirt1(Sir2) Antibody, Use Anti-Sirt1(Sir2) Antibody (Rabbit Polyclonal Antibody) is published (more than 30 citations) and validated in ICC, IP, WB to detect Sirt1(Sir2) also known as Silencing information regulator 2-like, sirtuin 2.
HPA000440
Anti-CD45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22763 Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500247
Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405848
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701826
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701825
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103104
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4300645
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody
HPA040067
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21465 Anti-PTPRC antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
PLA0302
Goat anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB4700471 Monoclonal Anti-CD45 antibody produced in mouse, clone MEM-28, purified immunoglobulin, buffered aqueous solution
AMAB90518
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0159, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90519
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0160, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700587 Monoclonal Anti-CD45 antibody produced in rat, clone EM-05, purified immunoglobulin, buffered aqueous solution
WH0002597M1
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
SAB4700649 Monoclonal Anti-PTPRC antibody produced in mouse, clone IVA103, purified immunoglobulin, buffered aqueous solution
PLA0125
Rabbit anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
252433
Rhodamine 6G, Dye content 99 %
C28H30N2O3 · HCl
56226
Rhodamine 6G, analytical standard
C28H30N2O3 · HCl
R4127
Rhodamine 6G, Dye content ~95 %
C28H30N2O3 · HCl
83697
Rhodamine 6G, suitable for fluorescence, BioReagent
C28H30N2O3 · HCl