EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Effect of high-intensity exercise on aged mouse brain mitochondria, neurogenesis, and inflammation.
PMID 25002036
Abstract
In aged mice, we assessed how intensive exercise affects brain bioenergetics, inflammation, and neurogenesis-relevant parameters. After 8 weeks of a supra-lactate threshold treadmill exercise intervention, 21-month-old C57BL/6 mice showed increased brain peroxisome proliferator-activated receptor gamma coactivator-1α protein, mammalian target of rapamycin and phospho-mammalian target of rapamycin protein, citrate synthase messenger RNA, and mitochondrial DNA copy number. Hippocampal vascular endothelial growth factor A (VEGF-A) gene expression trended higher, and a positive correlation between VEGF-A and PRC messenger RNA levels was observed. Brain doublecortin, brain-derived neurotrophic factor, tumor necrosis factor-α, and CCL11 gene expression, as well as plasma CCL11 protein levels, were unchanged. Despite these apparent negative findings, a negative correlation between plasma CCL11 protein levels and hippocampal doublecortin gene expression was observed; further analysis indicated exercise may mitigate this relationship. Overall, our data suggest supra-lactate threshold exercise activates a partial mitochondrial biogenesis in aged mice, and a gene (VEGF-A) known to support neurogenesis. Our data are consistent with another study that found systemic inflammation in general, and CCL11 protein specifically, suppresses hippocampal neurogenesis. Our study supports the view that intense exercise above the lactate threshold may benefit the aging brain; future studies to address the extent to which exercise-generated lactate mediates the observed effects are warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500268
Anti-COX4I1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405644
Anti-COX4I1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701802
Anti-cox4i1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA002485
Anti-COX4I1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401897
Anti-COX4I2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2104006
Anti-NFE2L1 antibody produced in rabbit, affinity isolated antibody
SAB2103657
Anti-NFE2L1 antibody produced in rabbit, affinity isolated antibody
SAB2701177
Anti-NFE2L1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2101653
Anti-NRF1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101654
Anti-NRF1 (ab2) antibody produced in rabbit, affinity isolated antibody
AV38543
Anti-NRF1 antibody produced in rabbit, IgG fraction of antiserum
HPA029329
Anti-NRF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
RAB0042
Human Eotaxin / CCL11 ELISA Kit, for serum, plasma, cell culture supernatant and urine
SAB5300321
Monoclonal Anti-COX4I1 antibody produced in mouse, clone 6B3, ascites fluid
SAB1402152
Monoclonal Anti-COX4I1 antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution
WH0084701M1
Monoclonal Anti-COX4I2 antibody produced in mouse, clone 1F2, purified immunoglobulin, buffered aqueous solution
SAB1404147
Monoclonal Anti-NRF1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
RAB0043
Mouse Eotaxin ELISA Kit, for serum, plasma and cell culture supernatant