EMAIL THIS PAGE TO A FRIEND
Oncotarget
Transcriptomic analysis reveals inhibition of androgen receptor activity by AMPK in prostate cancer cells.
PMID 25003216
Abstract
Metabolic alterations contribute to prostate cancer development and progression; however, the role of the central metabolic regulator AMP-activated protein kinase (AMPK) remains controversial. The androgen receptor (AR), a key driver of prostate cancer, regulates prostate cancer cell metabolism by driving the expression of a network of metabolic genes and activates AMPK through increasing the expression of one of its upstream kinases. To more clearly define the role of AMPK in prostate cancer, we performed expression profiling following pharmacologic activation of this kinase. We found that genes down-regulated upon AMPK activation were over-expressed in prostate cancer, consistent with a tumour suppressive function of AMPK. Strikingly, we identified the AR as one of the most significantly enriched transcription factors mediating gene expression changes downstream of AMPK signalling in prostate cancer cells. Activation of AMPK inhibited AR transcriptional activity and reduced androgen-dependent expression of known AR target genes. Conversely, knock-down of AMPK increased AR activity. Modulation of AR expression could not explain these effects. Instead, we observed that activation of AMPK reduced nuclear localisation of the AR. We thus propose the presence of a negative feedback loop in prostate cancer cells whereby AR activates AMPK and AMPK feeds back to limit AR-driven transcription.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001444
Bicalutamide, European Pharmacopoeia (EP) Reference Standard
C18H14F4N2O4S