EMAIL THIS PAGE TO A FRIEND
Shock (Augusta, Ga.)
Electroporation-mediated gene delivery of Na+,K+ -ATPase, and ENaC subunits to the lung attenuates acute respiratory distress syndrome in a two-hit porcine model.
PMID 25004064
Abstract
Acute respiratory distress syndrome (ARDS) is a common cause of organ failure with an associated mortality rate of 40%. The initiating event is disruption of alveolar-capillary interface causing leakage of edema into alveoli. Electroporation-mediated gene delivery of epithelial sodium channel (ENaC) and Na+,K+ -ATPase into alveolar cells would improve alveolar clearance of edema and attenuate ARDS. Pigs were anesthetized and instrumented, and the superior mesenteric artery was clamped to cause gut ischemia/reperfusion injury and peritoneal sepsis by fecal clot implantation. Animals were ventilated according to ARDSnet protocol. Four hours after injury, animals were randomized into groups: (i) treatment: Na+,K+ -ATPase/ENaC plasmid (n = 5) and (ii) control: empty plasmid (n = 5). Plasmids were delivered to the lung using bronchoscope. Electroporation was delivered using eight-square-wave electric pulses across the chest. Following electroporation, pigs were monitored 48 h. The Pao2/Fio2 ratio and lung compliance were higher in the treatment group. Lung wet/dry ratio was lower in the treatment group. Relative expression of the Na+,K+ -ATPase transgene was higher throughout lungs receiving treatment plasmids. Quantitative histopathology revealed a reduction in intra-alveolar fibrin in the treatment group. Bronchoalveolar lavage showed increased surfactant protein B in the treatment group. Survival was improved in the treatment group. Electroporation-mediated transfer of Na+,K+ -ATPase/ENaC plasmids improved lung function, reduced fibrin deposits, decreased lung edema, and improved survival in a translational porcine model of ARDS. Gene therapy can attenuate ARDS pathophysiology in a high-fidelity animal model, suggesting a potential new therapy for patients.