EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Shape dependent catalytic activity of nanoflowers and nanospheres of Pd4S generated via one pot synthesis and grafted on graphene oxide for Suzuki coupling.
PMID 25005160
Abstract
Nanoflowers and nanospheres of Pd4S have been prepared for the first time from a single source precursor complex, [PdCl2(PhS-CH2CH2CH2-NH2)] (), by its one pot thermolysis at 195 °C. In oleylamine, flower shaped nanoparticles of Pd4S were formed but in an oleic acid (OA) and octadecene (ODE) mixture (1 : 1) the product was nanospheres of Pd4S (size in the range ∼23-38 nm and 15-28 nm, respectively). These nanoparticles (NPs) were grafted on graphene oxide (GO) at room temperature to prepare nanocomposites, GO-Pd4S. HRTEM, powder X-ray diffraction (PXRD) and TEM-EDX have been used to authenticate the nanoparticles and their composites. XPS of Pd4S NPs indicates the oxidation states of Pd and S are both zero with a Pd : S ratio ∼4.1 : 0.9. For the catalysis of Suzuki-Miyaura coupling reactions the nanoparticles individually and in the form of composites with GO were explored. The flower shaped NPs are superior than the spherical ones for this catalysis in aqueous ethanol and the catalytic efficiency increases on grafting the nanoflowers/spheres onto GO. The conversion was ∼99% (in 5 h; at 80 °C) for the composite of graphene oxide (GO) with the Pd4S nanoflowers (Pd: 0.2 mol%). The catalytic efficiency follows the order GO-Pd4S-nanoflowers > GO-Pd4S-nanospheres > Pd4S nanoflowers > Pd4S nanospheres. The recyclability of the GO-Pd4S nanoflower catalyst was examined for the coupling reaction and conversion was found to be ∼46% in the fourth run even after increasing the reaction time to 12 h. To understand whether the catalytic process with the GO-Pd4S nanoflowers was homogeneous or heterogeneous mercury poisoning, triphenylphosphine and three phase tests were carried out. They suggest that active Pd leached from GO-Pd4S nanoflowers does the catalysis significantly in a homogeneous fashion. Overall the catalysis appears to be a cocktail of homogeneous and some heterogeneous nature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74738
1-Octadecene, analytical standard
C18H36
74740
1-Octadecene, ≥95.0% (GC)
C18H36
O806
1-Octadecene, technical grade, 90%
C18H36
74745
1-Octadecene, technical, ~85% (GC)
C18H36
142549
3-Chloropropylamine hydrochloride, 98%
C3H8ClN · HCl
255645
Cesium carbonate, 99.995% trace metals basis
CCs2O3
554855
Cesium carbonate, 99.95% trace metals basis
CCs2O3
202126
Cesium carbonate, 99.9% trace metals basis
CCs2O3
20959
Cesium carbonate, puriss. p.a., ≥99.0%
CCs2O3
441902
Cesium carbonate, ReagentPlus®, 99%
CCs2O3
20960
Cesium carbonate, purum p.a., ≥98.0% (T)
CCs2O3
O7805
Oleylamine, technical grade, 70%
C18H37N
HT-OA100
Oleylamine, ≥98% (primary amine)
C18H37N
78181
Phenylboronic acid, purum, ≥97.0% (HPLC)
C6H7BO2
P20009
Phenylboronic acid, 95%
C6H7BO2
791776
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
CK2O3
P5833
Potassium carbonate, BioXtra, ≥99.0%
CK2O3
P1472
Potassium carbonate, meets USP testing specifications
CK2O3
367877
Potassium carbonate, 99.995% trace metals basis
CK2O3
590681
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
CK2O3
60108
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
CK2O3
86857
Tetrabutylammonium bromide, for ion pair chromatography, ≥99.0%
C16H36BrN
426288
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
C16H36BrN
193119
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
C16H36BrN
462144
Tetrabutylammonium bromide solution, 50 wt. % in H2O
C16H36BrN
442272
1-Octadecene, analytical standard
C18H36