EMAIL THIS PAGE TO A FRIEND
eLife
Core promoter factor TAF9B regulates neuronal gene expression.
PMID 25006164
Abstract
Emerging evidence points to an unexpected diversification of core promoter recognition complexes that serve as important regulators of cell-type specific gene transcription. Here, we report that the orphan TBP-associated factor TAF9B is selectively up-regulated upon in vitro motor neuron differentiation, and is required for the transcriptional induction of specific neuronal genes, while dispensable for global gene expression in murine ES cells. TAF9B binds to both promoters and distal enhancers of neuronal genes, partially co-localizing at binding sites of OLIG2, a key activator of motor neuron differentiation. Surprisingly, in this neuronal context TAF9B becomes preferentially associated with PCAF rather than the canonical TFIID complex. Analysis of dissected spinal column from Taf9b KO mice confirmed that TAF9B also regulates neuronal gene transcription in vivo. Our findings suggest that alternative core promoter complexes may provide a key mechanism to lock in and maintain specific transcriptional programs in terminally differentiated cell types.DOI: http://dx.doi.org/10.7554/eLife.02559.001.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV40173
Anti-ACTB antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2100037
Anti-ACTB antibody produced in rabbit, affinity isolated antibody
GW23014 Anti-Actin, Cytoplasmic 1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500350
Anti-β-Actin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV100591
Anti-NANOG (AB1) antibody produced in rabbit, affinity isolated antibody
AV47988
Anti-NANOG (AB2) antibody produced in rabbit, affinity isolated antibody
SAB2500670
Anti-NANOG antibody produced in goat, affinity isolated antibody, buffered aqueous solution
N4413 Anti-Nanog antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4300680
Anti-Nanog antibody produced in rabbit, affinity isolated antibody
SAB3500389
Anti-NANOG antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104002
Anti-NANOG antibody produced in rabbit, affinity isolated antibody
N3038
Anti-Nanog antibody, Mouse monoclonal, clone NNG-811, purified from hybridoma cell culture
SAB2104004
Anti-NANOG, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1408519
Anti-NANOGP8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501671
Anti-NANOGP8 antibody produced in rabbit, affinity isolated antibody
SAB4502259
Anti-OCT3 antibody produced in rabbit, affinity isolated antibody
P7493
Anti-PCAF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV32466
Anti-PCAF antibody produced in rabbit, IgG fraction of antiserum
SAB2101734
Anti-PCAF antibody produced in rabbit, affinity isolated antibody
SAB2501454
Anti-POU5F1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV100956
Anti-POU5F1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101850
Anti-POU5F1 antibody produced in rabbit, affinity isolated antibody
SAB3500316
Anti-POU5F1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701958
Anti-POU5F1 antibody produced in rabbit
SAB2701960
Anti-POU5F1 antibody produced in rabbit
SAB2701972
Anti-POU5F1 antibody produced in rabbit
HPA004148
Anti-TAF10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV31426
Anti-TAF10 antibody produced in rabbit, affinity isolated antibody