PloS one
Moesin and stress-induced phosphoprotein-1 are possible sero-diagnostic markers of psoriasis.
PMID 25010044
Abstract
To identify diagnostic markers for psoriasis vulgaris and psoriatic arthritis, autoantibodies in sera from psoriasis vulgaris and psoriatic arthritis patients were screened by two-dimensional immunoblotting (2D-IB). Based on 2D-IB and MADLI TOF/TOF-MS analyses, eleven proteins each in psoriasis vulgaris and psoriatic arthritis were identified as autoantigens. Furthermore, serum levels of moesin, keratin 17 (K17), annexin A1 (ANXA1), and stress-induced phophoprotein-1 (STIP1), which were detected as autoantigens, were studied by dot blot analysis with psoriasis patients and healthy controls. The levels of moesin and STIP1 were significantly higher in sera from patients with psoriasis vulgaris than in the controls (moesin: P<0.05, STIP1: P<0.005). The area under the curve (AUC) for moesin and STIP1 between patients with psoraisis vulgaris and controls was 0.747 and 0.792, respectively. STIP1 and K17 levels were significantly higher in sera from patients with psoriatic arthritis than in those with psoriasis vulgaris (P<0.05 each). The AUC for STIP1 and K17 between patients with psoriatic arthritis and psoriasis vulgaris was 0.69 and 0.72, respectively. The STIP1 or moesin, CK17 serum level was not correlated with disease activity of psoriasis patients. These data suggest that STIP1 and moesin may be novel and differential sero-diagnostic markers for psoriasis vulgaris and psoriatic arthritis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA000452
Anti-JUP antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000453
Anti-JUP antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41733
Anti-KRT17 antibody produced in rabbit, affinity isolated antibody
SAB2500638
Anti-Moesin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA011135
Anti-MSN antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA011227
Anti-MSN antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107749
Anti-MSN antibody produced in rabbit, affinity isolated antibody
SAB2107173
Anti-MSN antibody produced in rabbit, affinity isolated antibody
SAB1410798
Anti-MSN antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV46164
Anti-STIP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV46165
Anti-STIP1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
GW22824 Anti-STIP1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407079
Anti-STIP1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA044062
Anti-STIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA039291
Anti-STIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0084525M1
Monoclonal Anti-HOP antibody produced in mouse, clone 3D6, purified immunoglobulin, buffered aqueous solution
WH0003872M1
Monoclonal Anti-KRT17 antibody produced in mouse, clone 2D4-1G9, purified immunoglobulin, buffered aqueous solution
SAB1400147
Monoclonal Anti-KRT17 antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
M7060
Monoclonal Anti-Moesin antibody produced in mouse, clone 38/87, purified immunoglobulin, buffered aqueous solution
WH0004478M13 Monoclonal Anti-MSN antibody produced in mouse, clone 4B8, purified immunoglobulin, buffered aqueous solution
SAB5300421
Monoclonal Anti-MSN antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
WH0010963M1
Monoclonal Anti-STIP1 antibody produced in mouse, clone 4B6, purified immunoglobulin, buffered aqueous solution
SAB1408802
Monoclonal Anti-STIP1 antibody produced in mouse, clone 2E1, purified immunoglobulin, buffered aqueous solution
SAB1408803
Monoclonal Anti-STIP1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
SAB1408804
Monoclonal Anti-STIP1 antibody produced in mouse, clone 1C6, purified immunoglobulin, buffered aqueous solution
SAB1408805
Monoclonal Anti-STIP1 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution