EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Unfolding and aggregation of a glycosylated monoclonal antibody on a cation exchange column. Part II. Protein structure effects by hydrogen deuterium exchange mass spectrometry.
PMID 25011681
Abstract
Hydrogen-deuterium exchange mass spectrometry (HX-MS) with proteolytic fragmentation is used to determine patterns of unfolding, as measured by increased solvent exposure, with peptide-level resolution for a glycosylated monoclonal antibody both when eluted from a tentacle-type cation exchange column (Fractogel EMD SO3-) and while bound to the resin. Two peaks are obtained when the bound protein is eluted with either a NaCl gradient or with two NaCl steps at increasing concentration. The first, early eluting peak contains only monomeric species whose structure is consistent with the native monomer. The second, late eluting peak contains a mixture of monomeric and aggregated species. The monomeric species in this mixture is also found to have a structure consistent with that of the native mAb, showing no evidence of increased solvent exposure. The aggregated species show instead significant unfolding in areas of the protein structure contained within the Fc region. The same peptides that exhibit the greatest level of solvent exposure in the aggregated species are also found in the fraction of protein that elutes from the resin only at high salt concentration, indicating that the aggregates are formed when the strongly-bound unfolded intermediate is desorbed at high salt. There is no evidence that the unfolded intermediate, formed while the protein is bound on the resin, is present in any of the eluted fractions indicating that, upon desorption from the resin, the intermediate either quickly refolds or forms aggregates which end-up co-eluting with the refolded protein at high salt concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59845
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
H8N2O4S
451975
Ammonium sulfate-14N2 solution, 99.99 atom % 14N, 40 wt. % in H2O
H814N2O4S
485411
Ammonium-14N2 sulfate, 99.99 atom % 14N
H814N2O4S
485306
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
H814N2O4S
608602
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
H814N216O4S
55561
Aucubin, analytical standard
C15H22O9
00090590
Aucubin, primary pharmaceutical reference standard
C15H22O9
38730 Citrate standard for IC, 1000 mg/L, analytical standard
C6H8O7
94676
Citric acid, certified reference material, TraceCERT®
C6H8O7
1134368
Citric acid, United States Pharmacopeia (USP) Reference Standard
C6H8O7
A1202000
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
C6H8O7
PHR1071
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
C6H8O7
633178
Deuterium oxide, 99.994 atom % D, contains 1 mM terephthalic acid disodium salt, 0.01 % (w/v) DSS-d6
D2O
530867
Deuterium oxide, 99.9 atom % D
D2O
613436
Deuterium oxide, 60 atom % D
D2O
661643
Deuterium oxide, 99.99 atom % D (for D2O), contains 1% DSS-d6
D2O
756822
Deuterium oxide, filtered, 99.8 atom % D
D2O
ARK2169
Ethanolamine, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for Pharma or biopharmaceutical production
C2H7NO
E9508
Ethanolamine, ≥98%
C2H7NO
02400
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
C2H7NO
E0135
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
C2H7NO
00278
Ethanolamine, analytical standard
C2H7NO
15014
Ethanolamine, ≥99%
C2H7NO
02412
Ethanolamine solution, for peptide synthesis, ~74% (T), ~70% in methanol
C2H7NO
42077
Sodium cyanoborohydride, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
NaBH3CN
156159
Sodium cyanoborohydride, reagent grade, 95%
NaBH3CN
71435
Sodium cyanoborohydride, purum, ≥95.0% (RT)
NaBH3CN
296813
Sodium cyanoborohydride solution, 1.0 M in THF
CH3BNNa
296945
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
CH3BNNa
746363
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Na2O4S