EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
IL-33 and thymic stromal lymphopoietin mediate immune pathology in response to chronic airborne allergen exposure.
PMID 25015831
Abstract
Humans are frequently exposed to various airborne allergens in the atmospheric environment. These allergens may trigger a complex network of immune responses in the airways, resulting in asthma and other chronic airway diseases. In this study, we investigated the immunological mechanisms involved in the pathological changes induced by chronic exposure to multiple airborne allergens. Naive mice were exposed intranasally to a combination of common airborne allergens, including the house dust mite, Alternaria, and Aspergillus, for up to 8 wk. These allergens acted synergistically and induced robust eosinophilic airway inflammation, specific IgE Ab production, type 2 cytokine response, and airway hyperresponsiveness in 4 wk, followed by airway remodeling in 8 wk. Increased lung infiltration of T cells, B cells, and type 2 innate lymphoid cells was observed. CD4(+) T cells and type 2 innate lymphoid cells contributed to the sources of IL-5 and IL-13, suggesting involvement of both innate and adaptive immunity in this model. The lung levels of IL-33 increased quickly within several hours after allergen exposure and continued to rise throughout the chronic phase of inflammation. Mice deficient in IL-33R (Il1rl1(-/-)) and thymic stromal lymphopoietin receptor (Tslpr(-/-)) showed significant reduction in airway inflammation, IgE Ab levels, and airway hyperresponsiveness. In contrast, mice deficient in IL-25R or IL-1R showed minimal differences as compared with wild-type animals. Thus, chronic exposure to natural airborne allergens triggers a network of innate and adaptive type 2 immune responses and airway pathology, and IL-33 and thymic stromal lymphopoietin most likely play key roles in this process.