EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Separation and characterization of phenolic compounds from dry-blanched peanut skins by liquid chromatography-electrospray ionization mass spectrometry.
PMID 25016324
Abstract
A large variety of phenolic compounds, including phenolic acids (hydroxybenzoic acids, hydroxycinnamic acids, and their esters), stilbenes (trans-resveratrol and trans-piceatannol), flavan-3-ols (e.g., (-)-epicatechin, (+)-catechin, and their polymers {the proanthocyanidins, PACs}), other flavonoids (e.g., isoflavones, flavanols, and flavones, etc.) and biflavonoids (e.g., morelloflavone), were identified in dry-blanched peanut skins (PS) by this study. High-performance liquid chromatography (HPLC) coupled with electrospray ionization mass spectrometry (ESI-MS(n)) was applied to separate and identify the phenolic constituents. Reversed-phase HPLC was employed to separate free phenolic compounds as well as PAC monomers, dimers, and trimers. PACs with a degree of polymerization (DP) of >4 were chromatographed via hydrophilic interaction liquid chromatography (HILIC). Tentative identification of the separated phenolics was based solely on molecular ions and MS(n) fragmentation patterns acquired by ESI-MS in the negative-ion mode. The connection sequence of PAC oligomers (DP <5) could be deduced mainly through characteristic quinone methide (QM) cleavage ions. When the DP reached 6, only a proportion of the flavan-3-ols could be ascertained in the PACs because of the extremely complicated fragmentation patterns involved. The identification of free phenolic acids, stilbenes, and flavonoids was achieved by authentic commercial standards and also by published literature data. Quantification was performed based on peak areas of the UV (free phenolic compounds) or fluorescence (PACs) signals from the HPLC chromatograms and calibration curves of commercial standards. Overall, PS contain significantly more PACs compared to free phenolic compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42157
Procyanidin B2, analytical standard
C30H26O12