EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Malarial pigment hemozoin impairs chemotactic motility and transendothelial migration of monocytes via 4-hydroxynonenal.
PMID 25017964
Abstract
Natural hemozoin, nHZ, is avidly phagocytosed in vivo and in vitro by human monocytes. The persistence of the undigested β-hematin core of nHZ in the phagocyte lysosome for long periods of time modifies several cellular immune functions. Here we show that nHZ phagocytosis by human primary monocytes severely impaired their chemotactic motility toward MCP-1, TNF, and FMLP, by approximately 80% each, and their diapedesis across a confluent human umbilical vein endothelial cell layer toward MCP-1 by 45±5%. No inhibition was observed with latex-fed or unfed monocytes. Microscopic inspection revealed polarization defects in nHZ-fed monocytes due to irregular actin polymerization. Phagocytosed nHZ catalyzes the peroxidation of polyunsaturated fatty acids and generation of the highly reactive derivative 4-hydroxynonenal (4-HNE). Similar to nHZ phagocytosis, the exposure of monocytes to in vivo-compatible 4-HNE concentrations inhibited cell motility in both the presence and the absence of chemotactic stimuli, suggesting severe impairment of cytoskeleton dynamics. Consequently, 4-HNE conjugates with the cytoskeleton proteins β-actin and coronin-1A were immunochemically identified in nHZ-fed monocytes and mass spectrometrically localized in domains of protein-protein interactions involved in cytoskeleton reorganization and cell motility. The molecular and functional modifications of actin and coronin by nHZ/4-HNE may also explain impaired phagocytosis, another motility-dependent process previously described in nHZ-fed monocytes. Further studies will show whether impaired monocyte motility may contribute to the immunodepression and the frequent occurrence of secondary infections observed in malaria patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22783 Anti-CD14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500215
Anti-CD14 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001887
Anti-CD14 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002127
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400126
Anti-HLA-E antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401182
Anti-HLA-E antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA031454
Anti-HLA-E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AMAB90897
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1637, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90898
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1638, purified immunoglobulin, buffered aqueous glycerol solution
C7673
Monoclonal Anti-CD14 antibody produced in mouse, clone UCHM-1, purified immunoglobulin, buffered aqueous solution
SAB4700097
Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
SAB4700103 Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700669 Monoclonal Anti-CD14 antibody produced in mouse, clone B-A8, purified immunoglobulin, buffered aqueous solution
SAB4700300 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/08, purified immunoglobulin, buffered aqueous solution
SAB4700301 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/06, purified immunoglobulin, buffered aqueous solution
SAB4700302 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/07, purified immunoglobulin, buffered aqueous solution