EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Capacity of the aquatic fern (Salvinia minima Baker) to accumulate high concentrations of nickel in its tissues, and its effect on plant physiological processes.
PMID 25019564
Abstract
An experiment was designed to assess the capacity of Salvinia minima Baker to uptake and accumulate nickel in its tissues and to evaluate whether or not this uptake can affect its physiology. Our results suggest that S. minima plants are able to take up high amounts of nickel in its tissues, particularly in roots. In fact, our results support the idea that S. minima might be considered a hyper-accumulator of nickel, as it is able to accumulate 16.3 mg g(-1) (whole plant DW basis). Our results also showed a two-steps uptake pattern of nickel, with a fast uptake of nickel at the first 6 to 12h of being expose to the metal, followed by a slow take up phase until the end of the experiment at 144 h. S. minima thus, may be considered as a fern useful in the phytoremediation of residual water bodies contaminated with this metal. Also from our results, S. minima can tolerate fair concentrations of the metal; however, at concentrations higher than 80 μM Ni (1.5 mg g(-1) internal nickel concentration), its physiological performance can be affected. For instance, the integrity of cell membranes was affected as the metal concentration and exposure time increased. The accumulation of high concentrations of internal nickel did also affect photosynthesis, the efficiency of PSII, and the concentration of photosynthetic pigments, although at a lower extent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF82156912
Nickel, rod, 100mm, diameter 12.0mm, 99%
Ni
GF11598194
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Ni
GF29576010
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Ni
GF83584143
Nickel, rod, 100mm, diameter 2.0mm, 99.99+%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF11517948
Nickel, rod, 100mm, diameter 20.0mm, 99%
Ni
GF39205931
Nickel, rod, 100mm, diameter 25.4mm, 99.98%
Ni
GF69167920
Nickel, rod, 100mm, diameter 3.0mm, 99%
Ni
GF70332870
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Ni
GF99799968
Nickel, rod, 100mm, diameter 30mm, 99%
Ni
GF90754241
Nickel, rod, 100mm, diameter 40mm, 99%
Ni
GF20561385
Nickel, rod, 100mm, diameter 5.0mm, 99.99+%
Ni
GF90932823
Nickel, rod, 100mm, diameter 6.35mm, 99%
Ni
GF50593002
Nickel, rod, 100mm, diameter 9.5mm, 99%
Ni
GF68355085
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Ni
GF66250256
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Ni
GF69613825
Nickel, rod, 200mm, diameter 12.0mm, 99%
Ni
GF49774034
Nickel, rod, 200mm, diameter 12.7mm, 99.98%
Ni
GF97332554
Nickel, rod, 200mm, diameter 16mm, 99.99+%
Ni
GF58660746
Nickel, rod, 200mm, diameter 2.0mm, 99.99+%
Ni
GF71556958
Nickel, rod, 200mm, diameter 2.0mm, annealed, 99%
Ni
GF39742577
Nickel, rod, 200mm, diameter 20.0mm, 99%
Ni
GF11244259
Nickel, rod, 200mm, diameter 25.4mm, 99.98%
Ni
GF51944665
Nickel, rod, 200mm, diameter 3.0mm, 99%
Ni
GF91609555
Nickel, rod, 200mm, diameter 3.3mm, 99.99+%
Ni
GF95866824
Nickel, rod, 200mm, diameter 30mm, 99%
Ni
GF44280974
Nickel, rod, 200mm, diameter 40mm, 99%
Ni
GF70172838
Nickel, rod, 200mm, diameter 5.0mm, 99.99+%
Ni
GF17475903
Nickel, rod, 200mm, diameter 6.35mm, 99%
Ni
GF24125071
Nickel, rod, 200mm, diameter 9.5mm, 99%
Ni