EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Inhibition of Hepatitis B virus replication by SAMHD1.
PMID 25019997
Abstract
Sterile alpha motif and HD-domain containing protein 1 (SAMHD1) is a newly identified intracellular antiviral factor. By depleting the dNTPs pool of host cells to a low level that cannot support the efficient synthesis of viral cDNA, it restricts replication of some retroviruses. As a DNA virus, Hepatitis B virus (HBV) experiences a process of reverse transcription in its life cycle akin to that of retroviruses. However, whether SAMHD1 can restrict HBV replication in liver cells is unknown. Here, we reported that SAMHD1 expression was detectable in four liver cell lines. Exogenous expression of SAMHD1 in SMMC-7721 cells restrained HBV replication. Similarly, SAMHD1 impeded HBV replication in another liver cell line, BEL-7402. Remarkably, the catalytically inactive mutant, SAMHD1 HD/AA also hampered HBV replication. Additionally, HBV replication reduced SAMHD1 expression in HepG2 cells. Moreover, it was found that IFN-α induced expression of SAMHD1 in liver cells. Together, these findings suggested that IFN-α-inducible SAMHD1 inhibited HBV replication in liver cells.