EMAIL THIS PAGE TO A FRIEND
Cellular signalling
Ret finger protein mediates Pax7-induced ubiquitination of MyoD in skeletal muscle atrophy.
PMID 25025573
Abstract
Skeletal muscle atrophy results from the net loss of muscular proteins and organelles and is caused by pathologic conditions such as nerve injury, immobilization, cancer, and other metabolic diseases. Recently, ubiquitination-mediated degradation of skeletal-muscle-specific transcription factors was shown to be involved in muscle atrophy, although the mechanisms have yet to be defined. Here we report that ret finger protein (RFP), also known as TRIM27, works as an E3 ligase in Pax7-induced degradation of MyoD. Muscle injury induced by sciatic nerve transection up-regulated RFP and RFP physically interacted with both Pax7 and MyoD. RFP and Pax7 synergistically reduced the protein amounts of MyoD but not the mRNA. RFP-induced reduction of MyoD protein was blocked by proteasome inhibitors. The Pax7-induced reduction MyoD was attenuated by RFP siRNA and by MG132, a proteasome inhibitor. RFPΔR, an RFP construct that lacks the RING domain, failed to reduce MyoD amounts. RFP ubiquitinated MyoD, but RFPΔR failed to do so. Forced expression of RFP, but not RFPΔR, enhanced Pax7-induced ubiquitination of MyoD, whereas RFP siRNA blocked the ubiquitination. Sciatic nerve injury-induced muscle atrophy as well the reduction in MyoD was attenuated in RFP knockout mice. Taken together, our results show that RFP works as a novel E3 ligase in the Pax7-mediated degradation of MyoD in response to skeletal muscle atrophy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501208
Anti-FBXO32 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1408235
Anti-FBXO32 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2106997
Anti-FBXO32 antibody produced in rabbit, affinity isolated antibody
SAB4300397
Anti-MYOD1 (Ab-200) antibody produced in rabbit, affinity isolated antibody
SAB2105139
Anti-MYOD1 antibody produced in rabbit, affinity isolated antibody
SAB1410813
Anti-MYOD1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV32393
Anti-PAX7 (AB1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV32742
Anti-PAX7 (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1412356
ANTI-PAX7 antibody produced in mouse, clone 1G11, purified immunoglobulin, buffered aqueous solution
SAB1412376
ANTI-PAX7 antibody produced in mouse, clone 3H1, purified immunoglobulin, buffered aqueous solution
SAB1412455
ANTI-PAX7 antibody produced in mouse, clone 4F8, purified immunoglobulin, buffered aqueous solution
SAB1412599
ANTI-PAX7 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
SAB1412600
ANTI-PAX7 antibody produced in mouse, clone 3F10, purified immunoglobulin, buffered aqueous solution
SAB1412601
ANTI-PAX7 antibody produced in mouse, clone 4F5, purified immunoglobulin, buffered aqueous solution
SAB2101730
Anti-PAX7 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2103961
Anti-PAX7 antibody produced in rabbit, affinity isolated antibody
EHU023151 MISSION® esiRNA, esiRNA human PAX7 (esiRNA1)
SAB1403439 Monoclonal Anti-FBXO32 antibody produced in mouse, clone 1D6, purified immunoglobulin, buffered aqueous solution
M6190 Monoclonal Anti-MYOD1 antibody produced in mouse, clone 5.2F, purified immunoglobulin, buffered aqueous solution
WH0004654M1
Monoclonal Anti-MYOD1 antibody produced in mouse, clone 1A7, purified immunoglobulin, buffered aqueous solution
SAB5300415
Monoclonal Anti-MYOD1 antibody produced in mouse, clone 1C8, ascites fluid
WH0005081M3
Monoclonal Anti-PAX7 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution