EMAIL THIS PAGE TO A FRIEND
Oncotarget
MRD assessed by WT1 and NPM1 transcript levels identifies distinct outcomes in AML patients and is influenced by gemtuzumab ozogamicin.
PMID 25026287
Abstract
We analysed the prognostic significance of minimal residual disease (MRD) level in adult patients with acute myeloid leukemia (AML) treated in the randomized gemtuzumab ozogamicin (GO) ALFA-0701 trial. Levels of WT1 and NPM1 gene transcripts were assessed using cDNA-based real-time quantitative PCR in 183 patients with WT1 overexpression and in 77 patients with NMP1 mutation (NPM1mut) at diagnosis. Positive WT1 MRD (defined as > 0.5% in the peripheral blood) after induction and at the end of treatment were both significantly associated with a higher risk of relapse and a shorter overall survival (OS). Positive NPM1mut MRD (defined as > 0.1% in the bone marrow) after induction and at the end of treatment also predicted a higher risk of relapse, but did not influence OS. Interestingly, the achievement of a negative NPM1mut MRD was significantly more frequent in patients treated in the GO arm compared to those treated in control arm (39 % versus 7% (p=0.006) after induction and 91% versus 61% (p=0.028) at the end of treatment). However, GO did not influence WT1 MRD levels. Our study supports the prognostic significance of MRD assessed by WT1 and NPM1mut transcript levels and show that NPM1 MRD is decreased by GO treatment.