EMAIL THIS PAGE TO A FRIEND
Archives of dermatological research
Calcipotriol counteracts betamethasone-induced decrease in extracellular matrix components related to skin atrophy.
PMID 25027750
Abstract
The calcipotriol/betamethasone dipropionate fixed-combination gel is widely used for topical treatment of psoriasis vulgaris. It has been hypothesized that calcipotriol counteracts glucocorticoid-induced skin atrophy which is associated with changes in the extracellular matrix (ECM). To elucidate the combined effects of calcipotriol and betamethasone on key ECM components, a comparative study to the respective mono-treatments was carried out. The effect on collagen I synthesis, matrix metalloproteinase (MMP) secretion, and hyaluronic acid (HA) production was investigated in primary human fibroblast and keratinocyte cultures as well as in a human skin explant model. We show that calcipotriol counteracts betamethasone-induced suppression of collagen I synthesis. Similarly, calcipotriol and betamethasone have opposing effects on MMP expression in both fibroblasts and keratinocytes. Moreover, calcipotriol is able to restore betamethasone-impaired HA synthesis in keratinocytes and prevent betamethasone-induced epidermal thinning in minipigs upon treatment with the calcipotriol/betamethasone gel. In summary, our results show for the first time in primary human skin cultures that calcipotriol reduces early signs of betamethasone-induced skin atrophy by modulation of key ECM components. These results indicate that the calcipotriol component of the fixed-combination gel counteracts the atrophogenic effects of betamethasone on the skin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB1912
Anti-Procollagen Type I Antibody, NT, clone M-58, 100 µL, Detect Procollagen Type I using this Anti-Procollagen Type I Antibody, N-terminus, clone M-58 validated for use in IH(P).
SAB1302705
ANTI-HAS1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2107778
Anti-HAS1 antibody produced in rabbit, affinity isolated antibody
SAB4501893
Anti-MMP3 (Cleaved-Phe100) antibody produced in rabbit, affinity isolated antibody
SAB1406132
Anti-MMP3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007875
Anti-MMP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
1067704 Betamethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
C28H37FO7
B1040000 Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
C28H37FO7
PHR1399 Betamethasone Dipropionate, Pharmaceutical Secondary Standard; Certified Reference Material
C28H37FO7
SAB5300089
Monoclonal Anti-HAS1 antibody produced in mouse, clone 3E10, ascites fluid
SAB5300422
Monoclonal Anti-MMP3 antibody produced in mouse, clone 4F10, purified immunoglobulin, buffered aqueous solution
SAB1402273
Monoclonal Anti-MMP3 antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution