EMAIL THIS PAGE TO A FRIEND
Biochemical genetics
Paraoxonase 1 polymorphisms within a Mississippi USA population as possible biomarkers of enzyme activities associated with disease susceptibility.
PMID 25027835
Abstract
Paraoxonase (PON1) hydrolyzes paraoxon (PO) and diazoxon (DZO), active metabolites of insecticides parathion and diazinon. The PON1 gene has single nucleotide polymorphisms (SNPs) including a codon 192 arginine (R) to glutamine (Q) and methionine (M) to leucine (L) at codon 55. Hydrolysis of PO (POase), DZO (DZOase), dihydrocoumarin (lactonase), and phenyl acetate (arylesterase) were evaluated for associations with race, gender, age, and PON1 55/192 SNP genotypes. Variables were analyzed both individually and in combination. QQ individuals had higher lactonase (pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9286
Paraoxon-ethyl, ≥90%, oil
C10H14NO6P
36186
Paraoxon-ethyl, PESTANAL®, analytical standard
C10H14NO6P