EMAIL THIS PAGE TO A FRIEND
Drug metabolism and pharmacokinetics
Inhibitory effects of calf thymus DNA on metabolism activity of CYP450 enzyme in human liver microsomes.
PMID 25030415
Abstract
The present study investigated the effect of calf thymus DNA (ctDNA) on human hepatic cytochrome P450s (CYP450s) in vitro. Specific substrate probes for each isoform, CYP1A2, 2C9, 2C19, 2D6 and 3A4, were incubated using pooled human liver microsomes with or without ctDNA, and liquid chromatography coupled tandem mass spectrometry (LC-MS/MS) method was developed for the analysis of probe metabolites. Enzyme kinetics parameters Ki and IC50 values were estimated to determine the types and strength of inhibition. ctDNA could specifically inhibit the metabolism of CYP2C9 probe substrates, with the IC50 = 0.9955 µg/ml, while it was not able to inhibit CYP1A2, CYP2C19, CYP2D6 or CYP3A4 (IC50 > 100 µg/ml). The results showed that ctDNA was a potent inhibitor of CYP2C9 enzyme, and has the metabolic interaction potential with the model drugs which are metabolism substrates of CYP2C9. The inhibition mechanism study suggested ctDNA may inhibit CYP2C9 by decreasing the activity of CYP450 reductase. These findings indicated that when the medical agents catalyzed mainly by CYP2C9 were co-administered in vivo with adsorptive material in vitro, the potential inhibitory effect of ctDNA on enzyme activity and the following metabolism character changes of the former should be highly focused on.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC126
4-Hydroxymephenytoin
C12H14N2O3
H146
4-Hydroxymephenytoin, ≥98% (HPLC)
C12H14N2O3
UC205
Dextrorphan
C17H23NO