EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
Effects of WT1 down-regulation on oocyte maturation and preimplantation embryo development in pigs.
PMID 25030893
Abstract
The Wilms' tumour 1 (WT1) gene originally identified as a tumour suppressor associated with WTs encodes a zinc finger-containing transcription factor that is expressed in multiple tissues and is an important regulator of cellular and organ growth, proliferation, development, migration and survival. However, there is a deficiency of data regarding the expression and function of WT1 during oocyte maturation and preimplantation embryonic development. Herein, we sought to define the expression characteristics and functions of WT1 during oocyte maturation and preimplantation embryonic development in pigs. We show that WT1 is expressed in porcine oocytes and at all preimplantation stages in embryos generated by ICSI. We then evaluated the effects of down-regulating WT1 expression at germinal vesicle and early ICSI stages using a recombinant plasmid (pGLV3-WT1-shRNA). Down-regulation of WT1 did not affect oocyte maturation but significantly decreased preimplantation embryonic development and increased apoptosis in blastocysts. These results indicate that WT1 plays important roles in the development of porcine preimplantation embryos.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700847 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700846 Anti-Rabbit IgG (Fc specific)-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700945 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700969 Anti-Rabbit IgG (H+L), F(ab) fragment-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
F1262 Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F6005 Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
SAB3700839 Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700828 Anti-Rabbit IgG F(ab′)2, highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700827 Anti-Rabbit IgG F(ab′)2-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
12-507
Goat Anti-Rabbit IgG (H+L) Antibody, FITC conjugate, Detect Rabbit IgG (H+L) using this Goat anti-Rabbit IgG (H+L) Antibody, FITC conjugate validated for use in FC, IF, IC, IH.
EHU083661 MISSION® esiRNA, esiRNA human WT1 (esiRNA1)