EMAIL THIS PAGE TO A FRIEND
Diabetologia
Fermentable fibres condition colon microbiota and promote diabetogenesis in NOD mice.
PMID 25031069
Abstract
Gut microbiota (GM) and diet both appear to be important in the pathogenesis of type 1 diabetes. Fermentable fibres (FFs), of which there is an ample supply in natural, diabetes-promoting diets, are used by GM as a source of energy. Our aim was to determine whether FFs modify GM and diabetes incidence in the NOD mouse. Female NOD mice were weaned to a semisynthetic diet and the effects of FF supplementation on diabetes incidence and insulitis were evaluated. Real-time quantitative PCR was employed to determine the effects imposed to gene transcripts in the colon and lymph nodes. Changes to GM were analysed by next-generation sequencing. NOD mice fed semisynthetic diets free from FFs were largely protected from diabetes while semisynthetic diets supplemented with the FFs pectin and xylan (PX) resulted in higher diabetes incidence. Semisynthetic diet free from FFs altered GM composition significantly; addition of PX changed the composition of the GM towards that found in natural-diet-fed mice and increased production of FF-derived short-chain fatty acid metabolites in the colon. The highly diabetogenic natural diet was associated with expression of proinflammatory and stress-related genes in the colon, while the semisynthetic diet free from FFs promoted Il4, Il22, Tgfβ and Foxp3 transcripts in the colon and/or pancreatic lymph node. PX in the same diet counteracted these effects and promoted stress-related IL-18 activation in gut epithelial cells. 16S RNA sequencing revealed each diet to give rise to its particular GM composition, with different Firmicutes to Bacteroidetes ratios, and enrichment of mucin-degrading Ruminococcaceae following diabetes-protective FF-free diet. FFs condition microbiota, affect colon homeostasis and are important components of natural, diabetes-promoting diets in NOD mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700555 Anti-Rat IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700541 Anti-Rat IgG (Fc specific)-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700548 Anti-Rat IgG (Fc specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700614 Anti-Rat IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700630 Anti-Rat IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700653 Anti-Rat IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700659 Anti-Rat IgG (H+L), F(ab) fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700606 Anti-Rat IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3700595 Anti-Rat IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700585 Anti-Rat IgG (H+L)-Peroxidase antibody produced in sheep, affinity isolated antibody, lyophilized powder
A9037 Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A5795 Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
A9542 Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3700533 Anti-Rat IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700526 Anti-Rat IgG F(ab′)2-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder