PloS one
Mechanisms of neuroblastoma cell growth inhibition by CARP-1 functional mimetics.
PMID 25033461
Abstract
Neuroblastomas (NBs) are a clinically heterogeneous group of extra cranial pediatric tumors. Patients with high-risk, metastatic NBs have a long-term survival rate of below 40%, and are often resistant to current therapeutic modalities. Due to toxic side effects associated with radiation and chemotherapies, development of new agents is warranted to overcome resistance and effectively treat this disease in clinic. CARP-1 functional mimetics (CFMs) are an emerging class of small molecule compounds that inhibit growth of diverse cancer cell types. Here we investigated NB inhibitory potential of CFMs and the molecular mechanisms involved. CFM-1, -4, and -5 inhibited NB cell growth, in vitro, independent of their p53 and MYCN status. CFM-4 and -5 induced apoptosis in NB cells in part by activating pro-apoptotic stress-activated kinases (SAPKs) p38 and JNK, stimulating CARP-1 expression and cleavage of PARP1, while promoting loss of the oncogenes C and N-myc as well as mitotic cyclin B1. Treatments of NB cells with CFM-4 or -5 also resulted in loss of Inhibitory κB (IκB) α and β proteins. Micro-RNA profiling revealed upregulation of XIAP-targeting miR513a-3p in CFM-4-treated NB, mesothelioma, and breast cancer cells. Moreover, exposure of NB and breast cancer cells to CFM-4 or -5 resulted in diminished expression of anti-apoptotic XIAP1, cIAP1, and Survivin proteins. Expression of anti-miR513a-5p or miR513a-5p mimic, however, interfered with or enhanced, respectively, the breast cancer cell growth inhibition by CFM-4. CFMs also impacted biological properties of the NB cells by blocking their abilities to migrate, form colonies in suspension, and invade through the matrix-coated membranes. Our studies indicate anti-NB properties of CFM-4 and 5, and suggest that these CFMs and/or their future analogs have potential as anti-NB agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405469
Anti-BIRC4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411890
ANTI-BIRC4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV30161
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100360
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB4503502
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB1409334
Anti-MYCN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410808
Anti-MYCN antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1412730
Anti-NEU1 antibody produced in mouse, purified immunoglobulin, 1 mg/mL
SAB4502972
Anti-TIMP2 antibody produced in rabbit, affinity isolated antibody
SAB2106577
Anti-TIMP2 antibody produced in rabbit, affinity isolated antibody
SAB2701028
Anti-TNIP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
X4503 Anti-XIAP antibody produced in goat, affinity isolated antibody, lyophilized powder
PRS3331
Anti-XIAP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501453
Anti-XIAP antibody produced in rabbit, affinity isolated antibody
WH0000331M1
Monoclonal Anti-BIRC4 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
SAB1403557
Monoclonal Anti-BIRC4 antibody produced in mouse, clone 1F4, purified immunoglobulin, buffered aqueous solution
WH0000891M1
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1C8, purified immunoglobulin, buffered aqueous solution
SAB5300329 Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1B10, ascites fluid
SAB5300330
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 5G6, ascites fluid
WH0004613M1
Monoclonal Anti-MYCN antibody produced in mouse, clone 3H4, purified immunoglobulin, buffered aqueous solution
SAB1404101
Monoclonal Anti-MYCN, (N-terminal) antibody produced in mouse, clone 4H4, purified immunoglobulin, buffered aqueous solution
SAB4100514
Monoclonal Anti-NEU1 antibody produced in mouse, clone NEU1-B591, culture supernatant
SAB1412731
Monoclonal Anti-NEU1 antibody produced in mouse, clone 3F9, purified immunoglobulin, 1 mg/mL
WH0007077M1
Monoclonal Anti-TIMP2 antibody produced in mouse, clone 5D7, purified immunoglobulin, buffered aqueous solution