EMAIL THIS PAGE TO A FRIEND
Restorative neurology and neuroscience
Forced limb-use enhances brain plasticity through the cAMP/PKA/CREB signal transduction pathway after stroke in adult rats.
PMID 25035316
Abstract
The mechanism underlying forced limb-use -induced structural plasticity remains to be studied. We examined whether the cyclic adenosine monophosphate (cAMP)-mediated signal transduction pathway was involved in brain plasticity and promoted behavioral recovery induced by forced limb-use after stroke. Adult rats were divided into a sham group, an ischemia group, an ischemia group with forced limb-use, and an ischemia group with forced limb-use and infusion of N-[2-(p-bromocinnamylamino)ethyl]-5-isoquinoline-sulfonamide (H89). Forced limb-use began on post-stroke day 7. Biotinylated dextran amine (BDA) was injected into the sensorimotor cortex on post-stroke day 14. Behavioral recovery was evaluated on post-stroke days 29 to 32, and the levels of cAMP, PKA C-α, phosphorylated CREB (pCREB), synaptophysin, PSD-95, BDA, and BrdU/NeuN were measured. The number of midline-crossing axons and the expression levels of synaptophysin and PSD-95 were increased after forced limb-use. Forced limb-use enhanced the survival of the newborn neurons and increased the levels of cAMP, PKA C-α and pCREB. These were significantly suppressed by H89. Behavioral performance improved with forced limb-use and was reversed with H89. Enhanced structural plasticity and the behavioral recovery promoted by post-stroke forced limb-use are suggested to be mediated through the cAMP/PKA/CREB signal transduction pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501245
Anti-DLG4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405717
Anti-DLG4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010122
Anti-DLG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401115
Anti-DLG4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105773
Anti-DLG4 antibody produced in rabbit, affinity isolated antibody
SAB4503908
Anti-phospho-CREB (pSer121) antibody produced in rabbit, affinity isolated antibody
C9102 Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4503757
Anti-phospho-CREB (pSer142) antibody produced in rabbit, affinity isolated antibody
SAB4504577
Anti-phospho-CREB (pThr100) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300217
Anti-phospho-CREB1 (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4502906
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
SAB4200544
Anti-Synaptophysin antibody, Mouse monoclonal, clone SVP-38, purified from hybridoma cell culture
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
SAB5200002
Monoclonal Anti-DLG4 antibody produced in mouse, clone 6G6, 1 mg/mL, purified immunoglobulin
SAB5200003
Monoclonal Anti-DLG4 antibody produced in mouse, clone 7E3, 1 mg/mL, purified immunoglobulin
P246
Monoclonal Anti-PSD95 antibody produced in mouse, clone 7E3-1B8, purified immunoglobulin, buffered aqueous solution
WH0006855M1
Monoclonal Anti-SYP antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution